Assay Details
| Gene Symbol: ZKSCAN8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278122.1 | NP_001265051.1 | 1 - 2 | 119 | 91 | |
| NM_006298.3 | NP_006289.2 | 1 - 2 | 119 | 91 | ||
| GenBank mRNA | AB208793.1 | - | 1 - 2 | 119 | 91 | |
| BC096283.3 | - | 1 - 2 | 91 | 40006903 | 91 | |
| BC096284.3 | - | 1 - 2 | 91 | 40006905 | 91 | |
| BC113961.1 | - | 1 - 2 | 128 | 40022943 | 91 | |
| BC130032.1 | - | 1 - 2 | 109 | 40018337 | 91 | |
| BC152378.1 | - | 1 - 2 | 128 | 40018341 | 91 | |
| U57796.1 | - | 1 - 2 | 91 | 91 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000276.hg
Additional Information:
This assay is in the 5'UTR region of the BC096283.3,BC096284.3,BC113961.1,BC130032.1,BC152378.1,NM_001278122.1,NM_006298.3,U57796.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06002288.hg.1 | ENST00000330236.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002288.hg.1 | ENST00000457389.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002288.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002288.hg.1 | TC06000276.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002292.hg.1 | ENST00000330236.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002292.hg.1 | ENST00000457389.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002292.hg.1 | ENST00000536028.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002292.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002292.hg.1 | TC06000276.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0600150092.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150092.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150092.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150092.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150092.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150092.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150092.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150092.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150092.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150092.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150092.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150092.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150092.hg.1 | TC0600007402.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600150097.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150097.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150097.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150097.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150097.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150097.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150097.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150097.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150097.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150097.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150097.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150097.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150097.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150097.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150097.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150097.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150097.hg.1 | TC0600007402.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150091.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150091.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150091.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150091.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150091.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150091.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150091.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150091.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150091.hg.1 | TC0600007402.hg.1 | Clariom D TC |

ZKSCAN8
Genomic Map