Assay Details
| Gene Symbol: ZFP57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109809.2 | NP_001103279.2 | 1 - 2 | 270 | 76 | |
| XM_011514570.2 | XP_011512872.1 | 2 - 3 | 571 | 76 | ||
| GenBank mRNA | BC157878.1 | - | 1 - 2 | 270 | 8862701 | 76 |
| BC171888.1 | - | 1 - 2 | 270 | 9054542 | 76 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001486.hg.1, TC6_apd_hap1000061.hg.1, TC6_cox_hap2000123.hg.1, TC6_dbb_hap3000112.hg.1, TC6_mann_hap4000101.hg.1, TC6_mcf_hap5000105.hg.1, TC6_qbl_hap6000113.hg.1, TC6_ssto_hap7000104.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002103.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002103.hg.1 | TC6_dbb_hap3000112.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001688.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001688.hg.1 | TC6_mann_hap4000101.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7001947.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7001947.hg.1 | TC6_ssto_hap7000104.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016701.hg.1 | ENST00000488757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016701.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016701.hg.1 | TC06001486.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002168.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002168.hg.1 | TC6_cox_hap2000123.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5001907.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5001907.hg.1 | TC6_mcf_hap5000105.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5001909.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5001909.hg.1 | TC6_mcf_hap5000105.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7001944.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7001944.hg.1 | TC6_ssto_hap7000104.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7001945.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7001945.hg.1 | TC6_ssto_hap7000104.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016699.hg.1 | ENST00000376881.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016699.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016699.hg.1 | ENST00000488757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016699.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016699.hg.1 | TC06001486.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001686.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001686.hg.1 | TC6_mann_hap4000101.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5001905.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5001905.hg.1 | TC6_mcf_hap5000105.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002139.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002139.hg.1 | TC6_qbl_hap6000113.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002140.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002140.hg.1 | TC6_qbl_hap6000113.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7001946.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7001946.hg.1 | TC6_ssto_hap7000104.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1000859.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1000859.hg.1 | TC6_apd_hap1000061.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002165.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002165.hg.1 | TC6_cox_hap2000123.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002102.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002102.hg.1 | TC6_dbb_hap3000112.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002104.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002104.hg.1 | TC6_dbb_hap3000112.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001685.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001685.hg.1 | TC6_mann_hap4000101.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5001906.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5001906.hg.1 | TC6_mcf_hap5000105.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002164.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002164.hg.1 | TC6_cox_hap2000123.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002100.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002100.hg.1 | TC6_dbb_hap3000112.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002101.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002101.hg.1 | TC6_dbb_hap3000112.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001687.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001687.hg.1 | TC6_mann_hap4000101.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5001908.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5001908.hg.1 | TC6_mcf_hap5000105.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7001943.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7001943.hg.1 | TC6_ssto_hap7000104.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001689.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001689.hg.1 | TC6_mann_hap4000101.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002141.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002141.hg.1 | TC6_qbl_hap6000113.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002142.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002142.hg.1 | TC6_qbl_hap6000113.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002167.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002167.hg.1 | TC6_cox_hap2000123.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002138.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002138.hg.1 | TC6_qbl_hap6000113.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011315.hg.1 | ENST00000376881.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011315.hg.1 | ENST00000376881.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011315.hg.1 | ENST00000376881.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011315.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011315.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011315.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011315.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011315.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011315.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011315.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011315.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011315.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011315.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011315.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011315.hg.1 | ENST00000488757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011315.hg.1 | ENST00000488757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011315.hg.1 | ENST00000488757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011315.hg.1 | ENST00000488757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011315.hg.1 | ENST00000488757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011315.hg.1 | ENST00000488757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011315.hg.1 | ENST00000488757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011315.hg.1 | ENST00000488757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011315.hg.1 | ENST00000488757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011315.hg.1 | ENST00000488757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002166.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002166.hg.1 | TC6_cox_hap2000123.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016700.hg.1 | ENST00000376881.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016700.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016700.hg.1 | ENST00000488757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016700.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016700.hg.1 | TC06001486.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016702.hg.1 | ENST00000376881.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016702.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016702.hg.1 | ENST00000488757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016702.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016702.hg.1 | TC06001486.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016703.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016703.hg.1 | ENST00000488757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016703.hg.1 | NM_001109809.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016703.hg.1 | TC06001486.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0600176243.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176243.hg.1 | ENST00000488757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176243.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176243.hg.1 | ENST00000488757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176243.hg.1 | TC0600011315.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600176244.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176244.hg.1 | ENST00000488757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176244.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176244.hg.1 | ENST00000488757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176244.hg.1 | TC0600011315.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600176239.hg.1 | ENST00000376881.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176239.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176239.hg.1 | ENST00000488757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176239.hg.1 | ENST00000376881.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176239.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176239.hg.1 | ENST00000488757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176239.hg.1 | TC0600011315.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600011315.hg.1 | ENST00000376881.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011315.hg.1 | ENST00000376881.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011315.hg.1 | ENST00000376881.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011315.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011315.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011315.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011315.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011315.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011315.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011315.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011315.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011315.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011315.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011315.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011315.hg.1 | ENST00000488757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011315.hg.1 | ENST00000488757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011315.hg.1 | ENST00000488757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011315.hg.1 | ENST00000488757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011315.hg.1 | ENST00000488757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011315.hg.1 | ENST00000488757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011315.hg.1 | ENST00000488757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011315.hg.1 | ENST00000488757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011315.hg.1 | ENST00000488757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011315.hg.1 | ENST00000488757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176241.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176241.hg.1 | ENST00000488757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176241.hg.1 | ENST00000376883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176241.hg.1 | ENST00000488757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176241.hg.1 | TC0600011315.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600176242.hg.1 | ENST00000488757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176242.hg.1 | ENST00000488757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176242.hg.1 | TC0600011315.hg.1 | Clariom D TC |

ZFP57
Genomic Map