Assay Details
| Gene Symbol: SETD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171990.2 | NP_001165461.1 | 2 - 3 | 847 | 148 | |
| NM_001323018.1 | NP_001309947.1 | 2 - 3 | 696 | 148 | ||
| NM_001323019.1 | NP_001309948.1 | 2 - 3 | 847 | 148 | ||
| NM_001323020.1 | NP_001309949.1 | 2 - 3 | 696 | 148 | ||
| NM_001323022.1 | NP_001309951.1 | 2 - 3 | 424 | 148 | ||
| NM_153706.3 | NP_714917.2 | 2 - 3 | 847 | 148 | ||
| XM_011543139.2 | XP_011541441.1 | 2 - 3 | 861 | 148 | ||
| XM_011543140.2 | XP_011541442.1 | 2 - 3 | 475 | 148 | ||
| GenBank mRNA | AK309152.1 | - | 2 - 3 | 755 | 148 | |
| BC036528.1 | - | 2 - 3 | 834 | 4827793 | 148 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001323022.1 transcripts.
Set Membership: |
|

SETD9
Genomic Map