Assay Details
| Gene Symbol: RPS27L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015920.3 | NP_057004.1 | 2 - 3 | 253 | 149 | |
| GenBank mRNA | AF070668.1 | - | 2 - 3 | 175 | 149 | |
| AK024591.1 | - | 2 - 3 | 188 | 149 | ||
| BC003667.1 | - | 2 - 3 | 185 | 3827362 | 149 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001540.hg
Additional Information:
This assay is in the 5'UTR region of the AF070668.1,BC003667.1,NM_015920.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
ribosomal small subunit assembly
DNA repair
rRNA processing
translation
activation of cysteine-type endopeptidase activity involved in apoptotic process
cellular response to DNA damage stimulus
mitotic G1 DNA damage checkpoint
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
positive regulation of translation
structural constituent of ribosome
protein binding
translation activator activity
cysteine-type endopeptidase activator activity involved in apoptotic process
poly(A) RNA binding
metal ion binding
ribosomal small subunit assembly
DNA repair
rRNA processing
translation
activation of cysteine-type endopeptidase activity involved in apoptotic process
cellular response to DNA damage stimulus
mitotic G1 DNA damage checkpoint
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
positive regulation of translation
structural constituent of ribosome
protein binding
translation activator activity
cysteine-type endopeptidase activator activity involved in apoptotic process
poly(A) RNA binding
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100014761.hg.1 | ENST00000418010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014761.hg.1 | ENST00000418010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000330964.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000330964.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000330964.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000330964.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000330964.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000330964.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000330964.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000330964.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000330964.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000411926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000411926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000411926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000411926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000411926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000439025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000439025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000439025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000439025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000455271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000455271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000455271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000455271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000455271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000462430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000462430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000462430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000462430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000462430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000482846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000482846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000482846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000482846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000482846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000482846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000482846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000482846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000482846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000482846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000559763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | ENST00000559763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | NM_015920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | NM_015920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | NM_015920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | NM_015920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | NM_015920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | NM_015920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009702.hg.1 | NM_015920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014479.hg.1 | ENST00000330964.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014479.hg.1 | ENST00000411926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014479.hg.1 | ENST00000439025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014479.hg.1 | ENST00000455271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014479.hg.1 | ENST00000462430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014479.hg.1 | ENST00000559763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014479.hg.1 | NM_015920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014479.hg.1 | TC15001540.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1500171583.hg.1 | ENST00000330964.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500171583.hg.1 | ENST00000411926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500171583.hg.1 | ENST00000439025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500171583.hg.1 | ENST00000455271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500171583.hg.1 | ENST00000462430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500171583.hg.1 | ENST00000482846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500171583.hg.1 | ENST00000559763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500171583.hg.1 | NM_015920.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500171583.hg.1 | TC1500009702.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500171583.hg.1 | ENST00000330964.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500171583.hg.1 | ENST00000411926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500171583.hg.1 | ENST00000439025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500171583.hg.1 | ENST00000455271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500171583.hg.1 | NM_015920.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500171583.hg.1 | ENST00000462430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500171583.hg.1 | ENST00000482846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500171583.hg.1 | ENST00000559763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000330964.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000330964.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000330964.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000330964.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000330964.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000330964.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000330964.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000330964.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000330964.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000411926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000411926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000411926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000411926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000411926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000439025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000439025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000439025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000439025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000455271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000455271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000455271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000455271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000455271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000462430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000462430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000462430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000462430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000462430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000482846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000482846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000482846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000482846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000482846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000482846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000482846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000482846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000482846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000482846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000559763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | ENST00000559763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | NM_015920.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | NM_015920.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | NM_015920.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | NM_015920.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | NM_015920.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | NM_015920.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009702.hg.1 | NM_015920.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014761.hg.1 | ENST00000418010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014761.hg.1 | ENST00000418010.1 | Ensembl |

RPS27L
Genomic Map