Assay Details
| Gene Symbol: VAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006373.3 | NP_006364.2 | 3 - 4 | 887 | 94 | |
| GenBank mRNA | AK097072.1 | - | 4 - 5 | 714 | 94 | |
| AK290460.1 | - | 3 - 4 | 863 | 94 | ||
| AK298536.1 | - | 3 - 4 | 597 | 94 | ||
| AK315838.1 | - | 3 - 4 | 451 | 94 | ||
| BC001913.1 | - | 3 - 4 | 756 | 94 | ||
| BC008725.2 | - | 3 - 4 | 811 | 3162543 | 94 | |
| BC014279.2 | - | 3 - 4 | 793 | 3834206 | 94 | |
| BC015041.2 | - | 3 - 4 | 825 | 3919901 | 94 | |
| BC090858.1 | - | 3 - 4 | 825 | 6528247 | 94 | |
| BT007369.1 | - | 3 - 4 | 767 | 94 | ||
| U18009.1 | - | 3 - 4 | 525 | 94 | ||
| U25779.1 | - | 3 - 4 | 492 | 94 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001552.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17020761.hg.1 | ENST00000355653.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020761.hg.1 | ENST00000420567.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020761.hg.1 | ENST00000542468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020761.hg.1 | NM_006373.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020761.hg.1 | TC17001552.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020762.hg.1 | ENST00000355653.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020762.hg.1 | ENST00000420567.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020762.hg.1 | ENST00000542468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020762.hg.1 | NM_006373.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020762.hg.1 | TC17001552.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000355653.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000355653.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000355653.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000355653.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000355653.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000355653.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000355653.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000355653.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000355653.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000355653.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000355653.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000355653.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000355653.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000355653.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000420567.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000420567.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000420567.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000420567.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000420567.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000420567.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000420567.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000420567.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000420567.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000420567.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000587062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000587062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000587062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000587062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000587062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000587147.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000587147.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000587147.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000587147.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000587173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000587173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000587173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000587173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000587173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000587173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000587173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000587173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000587173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000587173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000589709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000589709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000589709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000589709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000589709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000589709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000589828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000589828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000589828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000589828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000590924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000590924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000590924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000590924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000590924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000590924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000592388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000592388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | ENST00000592388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | NM_006373.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | NM_006373.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | NM_006373.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | NM_006373.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | NM_006373.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | NM_006373.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | NM_006373.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | NM_006373.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | NM_006373.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | NM_006373.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | NM_006373.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | NM_006373.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | NM_006373.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010762.hg.1 | NM_006373.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700185726.hg.1 | ENST00000355653.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185726.hg.1 | ENST00000420567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185726.hg.1 | ENST00000587147.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185726.hg.1 | ENST00000587173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185726.hg.1 | ENST00000589709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185726.hg.1 | ENST00000590924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185726.hg.1 | NM_006373.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700185726.hg.1 | TC1700010762.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700185726.hg.1 | ENST00000355653.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185726.hg.1 | ENST00000420567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185726.hg.1 | ENST00000587147.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185726.hg.1 | ENST00000587173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185726.hg.1 | ENST00000589709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185726.hg.1 | ENST00000590924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185726.hg.1 | NM_006373.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000355653.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000355653.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000355653.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000355653.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000355653.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000355653.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000355653.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000355653.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000355653.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000355653.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000355653.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000355653.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000355653.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000355653.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000420567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000420567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000420567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000420567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000420567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000420567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000420567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000420567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000420567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000420567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000587062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000587062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000587062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000587062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000587062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000587147.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000587147.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000587147.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000587147.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000587173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000587173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000587173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000587173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000587173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000587173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000587173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000587173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000587173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000587173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000589709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000589709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000589709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000589709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000589709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000589709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000589828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000589828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000589828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000589828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000590924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000590924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000590924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000590924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000590924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000590924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000592388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000592388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | ENST00000592388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | NM_006373.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | NM_006373.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | NM_006373.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | NM_006373.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | NM_006373.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | NM_006373.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | NM_006373.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | NM_006373.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | NM_006373.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | NM_006373.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | NM_006373.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | NM_006373.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | NM_006373.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010762.hg.1 | NM_006373.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700185728.hg.1 | ENST00000355653.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185728.hg.1 | ENST00000420567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185728.hg.1 | ENST00000587173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185728.hg.1 | ENST00000589709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185728.hg.1 | ENST00000590924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185728.hg.1 | NM_006373.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700185728.hg.1 | TC1700010762.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700185728.hg.1 | ENST00000355653.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185728.hg.1 | ENST00000420567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185728.hg.1 | ENST00000587173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185728.hg.1 | ENST00000589709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185728.hg.1 | ENST00000590924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185728.hg.1 | NM_006373.3 | Refseq |

VAT1
Genomic Map