Assay Details
| Gene Symbol: NCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145458.2 | NP_001138930.2 | 5 - 6 | 723 | 69 | |
| NM_001242357.2 | NP_001229286.1 | 4 - 5 | 438 | 69 | ||
| GenBank mRNA | AJ006121.1 | - | 5 - 6 | 692 | 69 | |
| AJ006123.1 | - | 4 - 5 | 407 | 69 | ||
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000601903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000601903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000601903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000601903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000601903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000601903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000601903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000601903.1 | Ensembl |

NCR1
Genomic Map