Assay Details
| Gene Symbol: OTUD6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286745.1 | NP_001273674.1 | 5 - 6 | 961 | 117 | |
| NM_016023.3 | NP_057107.3 | 4 - 5 | 821 | 117 | ||
| XM_011517129.2 | XP_011515431.2 | 5 - 6 | 848 | 117 | ||
| GenBank mRNA | AF151836.1 | - | 4 - 5 | 645 | 117 | |
| AK291646.1 | - | 4 - 5 | 653 | 117 | ||
| AK293843.1 | - | 5 - 6 | 832 | 117 | ||
| AL137441.1 | - | 3 - 4 | 434 | 117 | ||
| BC029760.1 | - | 4 - 5 | 667 | 4343258 | 117 | |
| BX647772.1 | - | 4 - 5 | 702 | 117 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000553.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08007582.hg.1 | ENST00000285420.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08007582.hg.1 | ENST00000404789.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08007582.hg.1 | NM_016023.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08007582.hg.1 | TC08000553.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000285420.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000285420.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000285420.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000285420.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000285420.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000285420.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000285420.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000285420.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000285420.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000285420.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000285420.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000285420.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000285420.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000404789.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000404789.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000404789.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000404789.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000404789.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000404789.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000404789.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000404789.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000522894.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000522894.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000522894.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000522894.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000522894.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000522894.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000522894.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000522894.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000522894.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000522894.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000522894.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000524027.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000524027.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000524027.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000524027.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000524027.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000524027.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000615618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000615618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000615618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000615618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000615618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000615618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000615618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000615618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000615618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000615618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000615618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000615618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000615618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000615618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000617869.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000617869.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000617869.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000617869.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000617869.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000617869.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000617869.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000617869.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000617869.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000617869.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000617869.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000617869.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | ENST00000617869.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | NM_001286745.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | NM_001286745.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | NM_001286745.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | NM_001286745.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | NM_001286745.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | NM_001286745.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | NM_001286745.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | NM_001286745.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | NM_001286745.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | NM_001286745.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | NM_001286745.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | NM_001286745.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | NM_001286745.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | NM_001286745.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | NM_016023.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | NM_016023.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | NM_016023.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | NM_016023.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | NM_016023.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | NM_016023.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | NM_016023.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | NM_016023.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | NM_016023.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | NM_016023.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | NM_016023.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | NM_016023.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008197.hg.1 | NM_016023.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000285420.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000285420.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000285420.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000285420.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000285420.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000285420.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000285420.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000285420.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000285420.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000285420.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000285420.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000285420.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000285420.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000404789.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000404789.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000404789.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000404789.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000404789.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000404789.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000404789.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000404789.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000522894.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000522894.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000522894.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000522894.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000522894.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000522894.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000522894.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000522894.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000522894.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000522894.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000522894.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000524027.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000524027.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000524027.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000524027.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000524027.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000524027.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000615618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000615618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000615618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000615618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000615618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000615618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000615618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000615618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000615618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000615618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000615618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000615618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000615618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000615618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000617869.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000617869.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000617869.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000617869.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000617869.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000617869.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000617869.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000617869.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000617869.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000617869.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000617869.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000617869.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | ENST00000617869.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | NM_001286745.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | NM_001286745.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | NM_001286745.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | NM_001286745.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | NM_001286745.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | NM_001286745.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | NM_001286745.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | NM_001286745.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | NM_001286745.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | NM_001286745.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | NM_001286745.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | NM_001286745.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | NM_001286745.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | NM_001286745.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | NM_016023.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | NM_016023.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | NM_016023.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | NM_016023.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | NM_016023.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | NM_016023.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | NM_016023.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | NM_016023.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | NM_016023.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | NM_016023.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | NM_016023.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | NM_016023.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008197.hg.1 | NM_016023.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800156306.hg.1 | ENST00000285420.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156306.hg.1 | ENST00000404789.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156306.hg.1 | ENST00000522894.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156306.hg.1 | ENST00000615618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156306.hg.1 | ENST00000617869.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156306.hg.1 | NM_001286745.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800156306.hg.1 | NM_016023.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800156306.hg.1 | ENST00000285420.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156306.hg.1 | ENST00000404789.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156306.hg.1 | ENST00000522894.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156306.hg.1 | ENST00000615618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156306.hg.1 | ENST00000617869.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156306.hg.1 | NM_001286745.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800156306.hg.1 | NM_016023.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800156306.hg.1 | TC0800008197.hg.1 | Clariom D TC |

OTUD6B
Genomic Map