Assay Details
| Gene Symbol: DCUN1D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173475.3 | NP_775746.1 | 1 - 2 | 156 | 84 | |
| GenBank mRNA | BC040442.1 | - | 1 - 2 | 136 | 5590225 | 84 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC040442.1,NM_173475.3 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

DCUN1D3
Genomic Map