Assay Details
| Gene Symbol: LRRC17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005824.2 | NP_005815.2 | 4 - 5 | 1372 | 114 | |
| GenBank mRNA | U32907.1 | - | 4 - 5 | 1264 | 114 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000668.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07010385.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010385.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010385.hg.1 | TC07000668.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000455453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000455453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000455453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000455453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000485478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000485478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000485478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000485478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000485478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000498487.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000498487.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000498487.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010383.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010383.hg.1 | ENST00000339431.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010383.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010383.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010383.hg.1 | TC07000668.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010381.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010381.hg.1 | ENST00000485478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010381.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010381.hg.1 | TC07000668.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010382.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010382.hg.1 | ENST00000339431.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010382.hg.1 | ENST00000485478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010382.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010382.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010382.hg.1 | TC07000668.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0700161218.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161218.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161218.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161218.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161218.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161218.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161218.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161218.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161218.hg.1 | TC0700008657.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700161217.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161217.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161217.hg.1 | ENST00000485478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161217.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161217.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161217.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161217.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161217.hg.1 | ENST00000485478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161217.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161217.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161217.hg.1 | TC0700008657.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700161220.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161220.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161220.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161220.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161220.hg.1 | TC0700008657.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000455453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000455453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000455453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000455453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000485478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000485478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000485478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000485478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000485478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000498487.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000498487.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000498487.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161215.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161215.hg.1 | ENST00000485478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161215.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161215.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161215.hg.1 | ENST00000485478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161215.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161215.hg.1 | TC0700008657.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700161216.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161216.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161216.hg.1 | ENST00000485478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161216.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161216.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161216.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161216.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161216.hg.1 | ENST00000485478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161216.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161216.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161216.hg.1 | TC0700008657.hg.1 | Clariom D TC |

LRRC17
Genomic Map