Assay Details
| Gene Symbol: RCOR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173587.3 | NP_775858.2 | 5 - 6 | 872 | 63 | |
| XM_005273932.3 | XP_005273989.1 | 5 - 6 | 589 | 63 | ||
| GenBank mRNA | AB593132.1 | - | 5 - 6 | 1215 | 63 | |
| BC023587.2 | - | 5 - 6 | 872 | 4762190 | 63 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001897.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11022378.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11022378.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11022378.hg.1 | TC11001897.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11022379.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11022379.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11022379.hg.1 | TC11001897.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008487.hg.1 | ENST00000548294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11022377.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11022377.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11022377.hg.1 | TC11001897.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | ENST00000473926.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | ENST00000473926.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | ENST00000489217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | ENST00000489217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | ENST00000489217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | ENST00000489217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | ENST00000489217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | ENST00000489217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | ENST00000489217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011141.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11022376.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11022376.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11022376.hg.1 | TC11001897.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11022380.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11022380.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11022380.hg.1 | TC11001897.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11022375.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11022375.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11022375.hg.1 | TC11001897.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100184926.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184926.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184926.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184926.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184926.hg.1 | TC1100011141.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200008487.hg.1 | ENST00000548294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184927.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184927.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184927.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184927.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184927.hg.1 | TC1100011141.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | ENST00000473926.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | ENST00000473926.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | ENST00000489217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | ENST00000489217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | ENST00000489217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | ENST00000489217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | ENST00000489217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | ENST00000489217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | ENST00000489217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011141.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184930.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184930.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184930.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184930.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184930.hg.1 | TC1100011141.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100184928.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184928.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184928.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184928.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184928.hg.1 | TC1100011141.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100184929.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184929.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184929.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184929.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184929.hg.1 | TC1100011141.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100184931.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184931.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184931.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184931.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184931.hg.1 | TC1100011141.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100184925.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184925.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184925.hg.1 | ENST00000301459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184925.hg.1 | NM_173587.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184925.hg.1 | TC1100011141.hg.1 | Clariom D TC |

RCOR2
Genomic Map