Assay Details
| Gene Symbol: ICAM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003259.3 | NP_003250.3 | 4 - 5 | 1025 | 102 | |
| XM_011528229.1 | XP_011526531.1 | 5 - 6 | 1097 | 102 | ||
| GenBank mRNA | AX746344.1 | - | 4 - 5 | 999 | 102 | |
| BC026338.1 | - | 4 - 5 | 1023 | 4814795 | 102 | |
| BC030132.2 | - | 2 - 3 | 624 | 102 | ||
| U72671.1 | - | 4 - 5 | 1025 | 102 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000176.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19002662.hg.1 | ENST00000221980.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002662.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002662.hg.1 | TC19000176.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000586004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000586004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000586004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000586480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000586480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000586480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000586480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000586480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000586480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000586480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000586480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000586480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000586480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000586480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000586480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000587398.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000587398.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000587398.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000587398.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000587398.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000588912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000588912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | ENST00000588912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006979.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000586004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000586004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000586004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000586480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000586480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000586480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000586480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000586480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000586480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000586480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000586480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000586480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000586480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000586480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000586480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000587398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000587398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000587398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000587398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000587398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000588912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000588912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | ENST00000588912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006979.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900149641.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149641.hg.1 | ENST00000586480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149641.hg.1 | NM_003259.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900149641.hg.1 | TC1900006979.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900149641.hg.1 | ENST00000221980.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149641.hg.1 | ENST00000586480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149641.hg.1 | NM_003259.3 | Refseq |

ICAM5
Genomic Map