Assay Details
| Gene Symbol: SBF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011530709.2 | XP_011529011.1 | 3 - 4 | 328 | 79 | |
| XM_017028905.1 | XP_016884394.1 | 3 - 4 | 327 | 79 | ||
| GenBank mRNA | AB209682.1 | - | 2 - 3 | 198 | 79 | |
| AK057985.1 | - | 4 - 5 | 335 | 79 | ||
| BI821356.1 | - | 3 - 4 | 468 | 79 | ||
| CR749672.1 | - | 1 - 2 | 126 | 79 | ||
More Information
Set Membership: |
|

SBF1
Genomic Map