Assay Details
| Gene Symbol: NTHL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002528.6 | NP_002519.1 | 2 - 3 | 397 | 62 | |
| XM_017023253.1 | XP_016878742.1 | 2 - 3 | 400 | 62 | ||
| GenBank mRNA | AB001575.1 | - | 2 - 3 | 361 | 62 | |
| BC000391.2 | - | 2 - 3 | 354 | 62 | ||
| BC003014.1 | - | 2 - 3 | 354 | 2821314 | 62 | |
| HY057336.1 | - | 2 - 3 | 374 | 62 | ||
| U79718.1 | - | 2 - 3 | 394 | 62 | ||
| U81285.1 | - | 2 - 3 | 359 | 62 | ||
| Y09687.1 | - | 2 - 3 | 348 | 62 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000772.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of defense response to virus by host
base-excision repair, AP site formation
nucleotide-excision repair, DNA incision, 5'-to lesion
depyrimidination
mitophagy in response to mitochondrial depolarization
xenophagy
oxidized pyrimidine nucleobase lesion DNA N-glycosylase activity
double-stranded DNA binding
DNA-(apurinic or apyrimidinic site) lyase activity
endonuclease activity
protein binding
oxidized purine nucleobase lesion DNA N-glycosylase activity
DNA N-glycosylase activity
metal ion binding
4 iron, 4 sulfur cluster binding
positive regulation of defense response to virus by host
base-excision repair, AP site formation
nucleotide-excision repair, DNA incision, 5'-to lesion
depyrimidination
mitophagy in response to mitochondrial depolarization
xenophagy
oxidized pyrimidine nucleobase lesion DNA N-glycosylase activity
double-stranded DNA binding
DNA-(apurinic or apyrimidinic site) lyase activity
endonuclease activity
protein binding
oxidized purine nucleobase lesion DNA N-glycosylase activity
DNA N-glycosylase activity
metal ion binding
4 iron, 4 sulfur cluster binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000219066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000219066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000219066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000219066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000219066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000219066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000219066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000219066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000219066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000219066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000219066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000219066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000561841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000561841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000561841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000561841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000561841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000561841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000561841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000561841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000561841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000561841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000561862.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000561862.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000561862.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000561862.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000561862.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000562120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000562120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000562120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000562120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000562951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000562951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000562951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000562951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000562951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000562951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000565406.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000565406.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000565406.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000565406.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000565406.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000565406.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000565406.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000566380.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000566380.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000566380.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000566380.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000566380.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000566380.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000566380.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000566380.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000566380.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000566380.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000567727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000567727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000567727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000567727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000567727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000567727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000568513.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000568513.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000568513.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000568513.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000568513.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000568513.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000568513.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000568513.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000568513.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000623977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000623977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000623977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | ENST00000623977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | NM_002528.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | NM_002528.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | NM_002528.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | NM_002528.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | NM_002528.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | NM_002528.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | NM_002528.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | NM_002528.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | NM_002528.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | NM_002528.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | NM_002528.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009085.hg.1 | NM_002528.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011284.hg.1 | ENST00000219066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011284.hg.1 | NM_002528.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011284.hg.1 | TC16000772.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600168993.hg.1 | ENST00000219066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600168993.hg.1 | ENST00000561841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600168993.hg.1 | ENST00000562120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600168993.hg.1 | ENST00000565406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600168993.hg.1 | ENST00000568513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600168993.hg.1 | NM_002528.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600168993.hg.1 | TC1600009085.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600168993.hg.1 | ENST00000219066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600168993.hg.1 | ENST00000561841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600168993.hg.1 | ENST00000562120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600168993.hg.1 | ENST00000565406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600168993.hg.1 | ENST00000568513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600168993.hg.1 | NM_002528.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600168995.hg.1 | ENST00000219066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600168995.hg.1 | ENST00000561841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600168995.hg.1 | ENST00000562120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600168995.hg.1 | ENST00000566380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600168995.hg.1 | NM_002528.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600168995.hg.1 | TC1600009085.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600168995.hg.1 | ENST00000219066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600168995.hg.1 | ENST00000561841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600168995.hg.1 | ENST00000562120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600168995.hg.1 | ENST00000566380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600168995.hg.1 | NM_002528.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000219066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000219066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000219066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000219066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000219066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000219066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000219066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000219066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000219066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000219066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000219066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000219066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000561841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000561841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000561841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000561841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000561841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000561841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000561841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000561841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000561841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000561841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000561862.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000561862.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000561862.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000561862.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000561862.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000562120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000562120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000562120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000562120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000562951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000562951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000562951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000562951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000562951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000562951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000565406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000565406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000565406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000565406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000565406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000565406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000565406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000566380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000566380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000566380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000566380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000566380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000566380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000566380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000566380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000566380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000566380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000567727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000567727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000567727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000567727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000567727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000567727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000568513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000568513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000568513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000568513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000568513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000568513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000568513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000568513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000568513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000623977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000623977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000623977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | ENST00000623977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | NM_002528.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | NM_002528.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | NM_002528.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | NM_002528.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | NM_002528.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | NM_002528.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | NM_002528.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | NM_002528.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | NM_002528.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | NM_002528.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | NM_002528.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009085.hg.1 | NM_002528.6 | Refseq |

NTHL1
Genomic Map