Assay Details
| Gene Symbol: POLR2G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002696.2 | NP_002687.1 | 5 - 6 | 509 | 65 | |
| GenBank mRNA | AK312131.1 | - | 5 - 6 | 468 | 65 | |
| BC112162.1 | - | 5 - 6 | 448 | 8327630 | 65 | |
| BC112164.1 | - | 5 - 6 | 448 | 8327632 | 65 | |
| U20659.1 | - | 5 - 6 | 427 | 65 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000563.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
nuclear-transcribed mRNA catabolic process, exonucleolytic
mRNA splicing, via spliceosome
transcription-coupled nucleotide-excision repair
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
7-methylguanosine mRNA capping
apoptotic process
fibroblast growth factor receptor signaling pathway
gene expression
gene silencing by RNA
somatic stem cell population maintenance
snRNA transcription from RNA polymerase II promoter
positive regulation of translational initiation
positive regulation of viral transcription
positive regulation of nuclear-transcribed mRNA poly(A) tail shortening
single-stranded DNA binding
single-stranded RNA binding
DNA-directed RNA polymerase activity
translation initiation factor binding
nuclear-transcribed mRNA catabolic process, exonucleolytic
mRNA splicing, via spliceosome
transcription-coupled nucleotide-excision repair
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
7-methylguanosine mRNA capping
apoptotic process
fibroblast growth factor receptor signaling pathway
gene expression
gene silencing by RNA
somatic stem cell population maintenance
snRNA transcription from RNA polymerase II promoter
positive regulation of translational initiation
positive regulation of viral transcription
positive regulation of nuclear-transcribed mRNA poly(A) tail shortening
single-stranded DNA binding
single-stranded RNA binding
DNA-directed RNA polymerase activity
translation initiation factor binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11006795.hg.1 | ENST00000301788.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006795.hg.1 | NM_002696.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006795.hg.1 | TC11000563.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000301788.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000301788.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000301788.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000301788.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000301788.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000301788.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000301788.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000524819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000524819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000524819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000524819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000524819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000524819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000524819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000524819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000525455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000525455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000525455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000525455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000525455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000525455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000525455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000525455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000526368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000526368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000526368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000526368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000526368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000526368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000526368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000527435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000527435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000527435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000527435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000527435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000527435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000527435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000527435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000531944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000531944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000531944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000531944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000531944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000531944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000531944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000531944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000531944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000531944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000531996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000531996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000531996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000531996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000531996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000531996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000533442.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000533442.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | ENST00000533442.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | NM_002696.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | NM_002696.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | NM_002696.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | NM_002696.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | NM_002696.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | NM_002696.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007874.hg.1 | NM_002696.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006794.hg.1 | ENST00000301788.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006794.hg.1 | NM_002696.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006794.hg.1 | TC11000563.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000301788.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000301788.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000301788.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000301788.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000301788.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000301788.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000301788.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000524819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000524819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000524819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000524819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000524819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000524819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000524819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000524819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000525455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000525455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000525455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000525455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000525455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000525455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000525455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000525455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000526368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000526368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000526368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000526368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000526368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000526368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000526368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000527435.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000527435.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000527435.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000527435.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000527435.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000527435.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000527435.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000527435.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000531944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000531944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000531944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000531944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000531944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000531944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000531944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000531944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000531944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000531944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000531996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000531996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000531996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000531996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000531996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000531996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000533442.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000533442.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | ENST00000533442.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | NM_002696.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | NM_002696.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | NM_002696.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | NM_002696.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | NM_002696.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | NM_002696.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007874.hg.1 | NM_002696.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100156074.hg.1 | ENST00000301788.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156074.hg.1 | ENST00000524819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156074.hg.1 | ENST00000525455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156074.hg.1 | ENST00000526368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156074.hg.1 | ENST00000527435.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156074.hg.1 | ENST00000531944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156074.hg.1 | ENST00000531996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156074.hg.1 | NM_002696.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100156074.hg.1 | ENST00000301788.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156074.hg.1 | ENST00000524819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156074.hg.1 | ENST00000525455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156074.hg.1 | ENST00000526368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156074.hg.1 | ENST00000527435.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156074.hg.1 | ENST00000531944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156074.hg.1 | ENST00000531996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156074.hg.1 | NM_002696.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100156074.hg.1 | TC1100007874.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100156075.hg.1 | ENST00000301788.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156075.hg.1 | ENST00000524819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156075.hg.1 | ENST00000525455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156075.hg.1 | ENST00000526368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156075.hg.1 | ENST00000527435.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156075.hg.1 | ENST00000531944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156075.hg.1 | ENST00000531996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156075.hg.1 | NM_002696.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100156075.hg.1 | ENST00000301788.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156075.hg.1 | ENST00000524819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156075.hg.1 | ENST00000525455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156075.hg.1 | ENST00000526368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156075.hg.1 | ENST00000527435.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156075.hg.1 | ENST00000531944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156075.hg.1 | ENST00000531996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156075.hg.1 | NM_002696.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100156075.hg.1 | TC1100007874.hg.1 | Clariom D TC |

POLR2G
Genomic Map