Assay Details
| Gene Symbol: WWC2-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024008.1 | 1 | 1675 | 96 | ||
| GenBank mRNA | AK054839.1 | - | 1 | 1675 | 96 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 119228 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 42045 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 407542 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P354496 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 18878 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 11450 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_14_P138241 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P354496 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_24_P354496 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837518 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676595 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837535 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837515 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837513 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676608 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837539 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837536 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676590 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676611 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676610 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837525 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837512 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837533 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837524 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676585 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676613 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676597 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676596 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837520 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676604 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676576 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676594 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837517 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676592 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837534 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676610 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676597 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676594 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676613 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676592 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837512 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837517 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676595 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837513 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837536 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837535 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837534 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676590 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676608 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837518 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676611 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837524 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837525 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837515 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676585 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676596 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676604 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837539 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837533 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676576 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837520 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P162128 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P354496 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P138241 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P354496 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676574 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676599 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676575 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676578 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15644675 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676571 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676588 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676606 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676590 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676592 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15644672 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676581 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676585 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676598 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676611 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15644669 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15644677 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676609 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676583 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676603 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P138241 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8103869 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8103870 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8103868 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8103871 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8103872 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8103867 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 230572_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1557685_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1557685_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 230572_at | - | - |
| Human Genome U133 Plus 2.0 Array | 230572_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557685_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230572_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557685_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230572_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230572_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557685_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557685_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557685_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557685_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557685_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230572_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557685_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230572_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557685_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230572_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230572_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557685_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230572_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230572_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230572_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557685_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230572_at2 | - | - |
| Human Genome U95 Set | 59015_PM_at | - | - |
| Human Genome U95 Set | 59015_at | - | - |
| Human Genome U133 Set | 230572_PM_at | - | - |
| Human Genome U133 Set | 230572_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A025052 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A025052 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 225150 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1757137 | AK054839.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1757137 | NR_024008.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1757137 | AK054839.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1757137 | NR_024008.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1757137 | AK054839.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1757137 | NR_024008.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1757137 | AK054839.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1757137 | NR_024008.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1757137 | AK054839.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1757137 | NR_024008.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1757137 | AK054839.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1757137 | NR_024008.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1757137 | AK054839.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1757137 | NR_024008.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1757137 | AK054839.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1757137 | NR_024008.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1757137 | AK054839.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1757137 | NR_024008.1 | human_transcriptome |

WWC2-AS2
Genomic Map