| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025450.hg.1 |
ENST00000230990.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025450.hg.1 |
ENST00000507104.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025450.hg.1 |
NM_001945.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025450.hg.1 |
TC05001854.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025449.hg.1 |
ENST00000230990.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025449.hg.1 |
ENST00000507104.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025449.hg.1 |
NM_001945.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025449.hg.1 |
TC05001854.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012238.hg.1 |
ENST00000230990.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012238.hg.1 |
ENST00000230990.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012238.hg.1 |
ENST00000230990.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012238.hg.1 |
ENST00000230990.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012238.hg.1 |
ENST00000230990.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012238.hg.1 |
ENST00000230990.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012238.hg.1 |
ENST00000230990.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012238.hg.1 |
ENST00000230990.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012238.hg.1 |
ENST00000230990.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012238.hg.1 |
ENST00000482211.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012238.hg.1 |
ENST00000482211.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012238.hg.1 |
ENST00000482211.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012238.hg.1 |
ENST00000498452.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012238.hg.1 |
ENST00000498452.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012238.hg.1 |
ENST00000498452.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012238.hg.1 |
NM_001945.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012238.hg.1 |
NM_001945.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012238.hg.1 |
NM_001945.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012238.hg.1 |
NM_001945.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012238.hg.1 |
NM_001945.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012238.hg.1 |
NM_001945.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012238.hg.1 |
NM_001945.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012238.hg.1 |
NM_001945.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025448.hg.1 |
ENST00000230990.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025448.hg.1 |
ENST00000507104.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025448.hg.1 |
NM_001945.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025448.hg.1 |
TC05001854.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0500184962.hg.1 |
ENST00000230990.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184962.hg.1 |
NM_001945.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500184962.hg.1 |
ENST00000230990.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184962.hg.1 |
NM_001945.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500184962.hg.1 |
TC0500012238.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500184963.hg.1 |
ENST00000230990.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184963.hg.1 |
NM_001945.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500184963.hg.1 |
ENST00000230990.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184963.hg.1 |
NM_001945.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500184963.hg.1 |
TC0500012238.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500012238.hg.1 |
ENST00000230990.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012238.hg.1 |
ENST00000230990.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012238.hg.1 |
ENST00000230990.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012238.hg.1 |
ENST00000230990.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012238.hg.1 |
ENST00000230990.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012238.hg.1 |
ENST00000230990.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012238.hg.1 |
ENST00000230990.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012238.hg.1 |
ENST00000230990.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012238.hg.1 |
ENST00000230990.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012238.hg.1 |
ENST00000482211.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012238.hg.1 |
ENST00000482211.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012238.hg.1 |
ENST00000482211.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012238.hg.1 |
ENST00000498452.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012238.hg.1 |
ENST00000498452.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012238.hg.1 |
ENST00000498452.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012238.hg.1 |
NM_001945.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012238.hg.1 |
NM_001945.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012238.hg.1 |
NM_001945.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012238.hg.1 |
NM_001945.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012238.hg.1 |
NM_001945.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012238.hg.1 |
NM_001945.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012238.hg.1 |
NM_001945.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012238.hg.1 |
NM_001945.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500184961.hg.1 |
ENST00000230990.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184961.hg.1 |
NM_001945.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500184961.hg.1 |
ENST00000230990.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184961.hg.1 |
NM_001945.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500184961.hg.1 |
TC0500012238.hg.1 |
Clariom D TC |