| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
8079421 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8079420 |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
236731_PM_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007264.hg.1 |
ENST00000599511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007264.hg.1 |
ENST00000599511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007264.hg.1 |
ENST00000599511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007264.hg.1 |
ENST00000599511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007264.hg.1 |
ENST00000599511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007264.hg.1 |
ENST00000614743.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007264.hg.1 |
ENST00000614743.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007264.hg.1 |
ENST00000614743.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007264.hg.1 |
NR_073385.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007264.hg.1 |
NR_073385.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007264.hg.1 |
NR_073385.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007264.hg.1 |
ENST00000599511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007264.hg.1 |
ENST00000599511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007264.hg.1 |
ENST00000599511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007264.hg.1 |
ENST00000599511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007264.hg.1 |
ENST00000599511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007264.hg.1 |
ENST00000614743.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007264.hg.1 |
ENST00000614743.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007264.hg.1 |
ENST00000614743.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007264.hg.1 |
NR_073385.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007264.hg.1 |
NR_073385.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007264.hg.1 |
NR_073385.1 |
Refseq |
| Human Genome U133 Plus 2.0 Array |
236731_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
236731_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
236731_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
236731_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
236731_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
236731_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
236731_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
236731_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
236731_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
236731_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
236731_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
236731_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
236731_at11 |
- |
- |
| Human Genome U95 Set |
72451_PM_at |
- |
- |
| Human Genome U95 Set |
72451_at |
- |
- |
| Human Genome U133 Set |
236731_at |
- |
- |
| Human Genome U133 Set |
236731_PM_at |
- |
- |