Assay Details
| Gene Symbol: STAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005843.5 | NP_005834.4 | 6 - 7 | 867 | 91 | |
| GenBank mRNA | AF042273.1 | - | 6 - 7 | 626 | 91 | |
| AF042274.1 | - | 6 - 7 | 626 | 91 | ||
| AK292265.1 | - | 6 - 7 | 562 | 91 | ||
| AK310863.1 | - | 6 - 7 | 604 | 91 | ||
| AL133600.1 | - | 6 - 7 | 629 | 91 | ||
| BC028740.2 | - | 6 - 7 | 867 | 5260998 | 91 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002432.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000465460.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000465460.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000465460.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000465460.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000465460.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000465460.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000465460.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000489389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000489389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000489389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000489389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000489389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000494589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000494589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000494589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000494589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000494589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000494589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | ENST00000494589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02038386.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02038386.hg.1 | ENST00000465460.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02038386.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02038386.hg.1 | TC02002432.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0200206440.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206440.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206440.hg.1 | ENST00000465460.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206440.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206440.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200206440.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206440.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206440.hg.1 | ENST00000465460.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206440.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206440.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200206440.hg.1 | TC0200014623.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000465460.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000465460.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000465460.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000465460.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000465460.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000465460.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000465460.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000489389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000489389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000489389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000489389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000489389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000494589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000494589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000494589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000494589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000494589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000494589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | ENST00000494589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014623.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200206441.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206441.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206441.hg.1 | ENST00000465460.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206441.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206441.hg.1 | ENST00000494589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206441.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200206441.hg.1 | ENST00000263904.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206441.hg.1 | ENST00000463854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206441.hg.1 | ENST00000465460.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206441.hg.1 | ENST00000482997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206441.hg.1 | ENST00000494589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206441.hg.1 | NM_005843.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200206441.hg.1 | TC0200014623.hg.1 | Clariom D TC |

STAM2
Genomic Map