| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05006068.hg.1 |
ENST00000274376.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05006068.hg.1 |
ENST00000456692.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05006068.hg.1 |
ENST00000506290.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05006068.hg.1 |
ENST00000512763.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05006068.hg.1 |
ENST00000534133.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05006068.hg.1 |
NM_002890.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05006068.hg.1 |
NM_022650.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05006068.hg.1 |
TC05000431.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0500154825.hg.1 |
ENST00000274376.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500154825.hg.1 |
ENST00000456692.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500154825.hg.1 |
ENST00000506290.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500154825.hg.1 |
ENST00000509953.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500154825.hg.1 |
ENST00000512763.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500154825.hg.1 |
ENST00000515800.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500154825.hg.1 |
NM_002890.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500154825.hg.1 |
NM_022650.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500154825.hg.1 |
ENST00000274376.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500154825.hg.1 |
ENST00000456692.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500154825.hg.1 |
ENST00000506290.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500154825.hg.1 |
ENST00000509953.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500154825.hg.1 |
ENST00000512763.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500154825.hg.1 |
ENST00000515800.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500154825.hg.1 |
NM_002890.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500154825.hg.1 |
NM_022650.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500154825.hg.1 |
TC0500008014.hg.1 |
Clariom D TC |