| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17018365.hg.1 |
ENST00000293280.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17018365.hg.1 |
NM_005064.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17018365.hg.1 |
TC17001384.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
ENST00000612516.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
ENST00000612516.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
ENST00000612516.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
ENST00000612516.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
ENST00000612516.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
ENST00000612516.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
ENST00000612516.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
ENST00000612516.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
ENST00000613876.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
ENST00000613876.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
ENST00000613876.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
ENST00000613876.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
ENST00000613876.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
ENST00000615050.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
ENST00000615050.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
ENST00000615050.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
ENST00000615050.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
ENST00000615050.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
ENST00000615050.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
ENST00000615050.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
ENST00000615050.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
ENST00000615050.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
NM_005064.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
NM_005064.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
NM_005064.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
NM_005064.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
NM_005064.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
NM_005064.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
NM_005064.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
NM_005064.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
NM_005064.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
NM_145898.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
NM_145898.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
NM_145898.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
NM_145898.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
NM_145898.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
NM_145898.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
NM_145898.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010456.hg.1 |
NM_145898.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700182101.hg.1 |
ENST00000615050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700182101.hg.1 |
NM_005064.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700182101.hg.1 |
TC1700010456.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700182101.hg.1 |
ENST00000615050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700182101.hg.1 |
NM_005064.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
ENST00000612516.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
ENST00000612516.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
ENST00000612516.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
ENST00000612516.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
ENST00000612516.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
ENST00000612516.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
ENST00000612516.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
ENST00000612516.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
ENST00000613876.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
ENST00000613876.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
ENST00000613876.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
ENST00000613876.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
ENST00000613876.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
ENST00000615050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
ENST00000615050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
ENST00000615050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
ENST00000615050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
ENST00000615050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
ENST00000615050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
ENST00000615050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
ENST00000615050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
ENST00000615050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
NM_005064.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
NM_005064.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
NM_005064.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
NM_005064.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
NM_005064.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
NM_005064.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
NM_005064.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
NM_005064.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
NM_005064.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
NM_145898.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
NM_145898.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
NM_145898.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
NM_145898.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
NM_145898.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
NM_145898.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
NM_145898.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010456.hg.1 |
NM_145898.3 |
Refseq |