| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
ENST00000259883.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
ENST00000259883.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
ENST00000259883.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
ENST00000259883.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
ENST00000259883.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
ENST00000259883.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
ENST00000259883.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
ENST00000259883.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
ENST00000259883.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
ENST00000259883.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
ENST00000259883.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014089.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002351.hg.1 |
ENST00000259883.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002351.hg.1 |
ENST00000405948.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002351.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002351.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002351.hg.1 |
TC06000282.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002347.hg.1 |
ENST00000405948.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002347.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002347.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002347.hg.1 |
TC06000282.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002348.hg.1 |
ENST00000259883.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002348.hg.1 |
ENST00000405948.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002348.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002348.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002348.hg.1 |
TC06000282.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002349.hg.1 |
ENST00000259883.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002349.hg.1 |
ENST00000405948.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002349.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002349.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002349.hg.1 |
TC06000282.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0600197829.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600197829.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600197829.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600197829.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600197829.hg.1 |
TC0600014089.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600197830.hg.1 |
ENST00000259883.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600197830.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600197830.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600197830.hg.1 |
ENST00000259883.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600197830.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600197830.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600197830.hg.1 |
TC0600014089.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
ENST00000259883.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
ENST00000259883.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
ENST00000259883.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
ENST00000259883.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
ENST00000259883.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
ENST00000259883.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
ENST00000259883.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
ENST00000259883.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
ENST00000259883.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
ENST00000259883.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
ENST00000259883.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014089.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600197833.hg.1 |
ENST00000259883.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600197833.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600197833.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600197833.hg.1 |
ENST00000259883.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600197833.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600197833.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600197833.hg.1 |
TC0600014089.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600197831.hg.1 |
ENST00000259883.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600197831.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600197831.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600197831.hg.1 |
ENST00000259883.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600197831.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600197831.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600197831.hg.1 |
TC0600014089.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600197834.hg.1 |
ENST00000259883.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600197834.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600197834.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600197834.hg.1 |
ENST00000259883.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600197834.hg.1 |
NM_001184743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600197834.hg.1 |
NM_032507.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600197834.hg.1 |
TC0600014089.hg.1 |
Clariom D TC |