Assay Details
| Gene Symbol: CHD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271.3 | NP_001262.3 | 27 - 28 | 4029 | 93 | |
| GenBank mRNA | AF006514.1 | - | 27 - 28 | 93 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15002769.hg.1
Set Membership: |
|

CHD2
Genomic Map