Assay Details
| Gene Symbol: PDGFA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033023.4 | NP_148983.1 | 5 - 6 | 1420 | 65 | |
| XM_011515417.1 | XP_011513719.1 | 6 - 7 | 912 | 65 | ||
| XM_011515418.1 | XP_011513720.1 | 6 - 7 | 916 | 65 | ||
| GenBank mRNA | A09204.1 | - | 5 - 6 | 613 | 65 | |
| X06374.1 | - | 5 - 6 | 980 | 65 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001073.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
angiogenesis
response to hypoxia
cell activation
hair follicle development
positive regulation of mesenchymal cell proliferation
platelet degranulation
transforming growth factor beta receptor signaling pathway
cell-cell signaling
positive regulation of cell proliferation
response to wounding
organ morphogenesis
response to inorganic substance
negative regulation of phosphatidylinositol biosynthetic process
negative regulation of platelet activation
regulation of phosphatidylinositol 3-kinase signaling
positive regulation of phosphatidylinositol 3-kinase signaling
regulation of smooth muscle cell migration
cell projection assembly
actin cytoskeleton organization
extracellular matrix organization
positive regulation of cell migration
positive regulation of protein autophosphorylation
response to estradiol
response to retinoic acid
regulation of actin cytoskeleton organization
positive regulation of metanephric mesenchymal cell migration by platelet-derived growth factor receptor-beta signaling pathway
wound healing
response to drug
positive regulation of MAP kinase activity
positive regulation of MAPK cascade
positive regulation of GTPase activity
skin development
positive regulation of DNA replication
phosphatidylinositol phosphorylation
platelet-derived growth factor receptor signaling pathway
phosphatidylinositol-mediated signaling
positive regulation of fibroblast proliferation
lung alveolus development
inner ear development
regulation of peptidyl-tyrosine phosphorylation
negative chemotaxis
positive regulation of cell division
positive regulation of protein kinase B signaling
regulation of branching involved in salivary gland morphogenesis by epithelial-mesenchymal signaling
positive regulation of ERK1 and ERK2 cascade
regulation of glomerular mesangial cell proliferation
embryonic lung development
regulation of DNA biosynthetic process
Ras guanyl-nucleotide exchange factor activity
platelet-derived growth factor receptor binding
protein binding
collagen binding
growth factor activity
protein homodimerization activity
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
protein heterodimerization activity
platelet-derived growth factor binding
MAPK cascade
angiogenesis
response to hypoxia
cell activation
hair follicle development
positive regulation of mesenchymal cell proliferation
platelet degranulation
transforming growth factor beta receptor signaling pathway
cell-cell signaling
positive regulation of cell proliferation
response to wounding
organ morphogenesis
response to inorganic substance
negative regulation of phosphatidylinositol biosynthetic process
negative regulation of platelet activation
regulation of phosphatidylinositol 3-kinase signaling
positive regulation of phosphatidylinositol 3-kinase signaling
regulation of smooth muscle cell migration
cell projection assembly
actin cytoskeleton organization
extracellular matrix organization
positive regulation of cell migration
positive regulation of protein autophosphorylation
response to estradiol
response to retinoic acid
regulation of actin cytoskeleton organization
positive regulation of metanephric mesenchymal cell migration by platelet-derived growth factor receptor-beta signaling pathway
wound healing
response to drug
positive regulation of MAP kinase activity
positive regulation of MAPK cascade
positive regulation of GTPase activity
skin development
positive regulation of DNA replication
phosphatidylinositol phosphorylation
platelet-derived growth factor receptor signaling pathway
phosphatidylinositol-mediated signaling
positive regulation of fibroblast proliferation
lung alveolus development
inner ear development
regulation of peptidyl-tyrosine phosphorylation
negative chemotaxis
positive regulation of cell division
positive regulation of protein kinase B signaling
regulation of branching involved in salivary gland morphogenesis by epithelial-mesenchymal signaling
positive regulation of ERK1 and ERK2 cascade
regulation of glomerular mesangial cell proliferation
embryonic lung development
regulation of DNA biosynthetic process
Ras guanyl-nucleotide exchange factor activity
platelet-derived growth factor receptor binding
protein binding
collagen binding
growth factor activity
protein homodimerization activity
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
protein heterodimerization activity
platelet-derived growth factor binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07016419.hg.1 | ENST00000402802.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07016419.hg.1 | NM_002607.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07016419.hg.1 | NM_033023.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07016419.hg.1 | TC07001073.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000354513.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000354513.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000354513.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000354513.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000354513.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000354513.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000354513.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000354513.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000354513.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000354513.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000400761.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000400761.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000400761.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000400761.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000400761.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000400761.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000402802.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000402802.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000402802.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000402802.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000402802.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000402802.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000402802.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000402802.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000402802.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000402802.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000405692.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000405692.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000405692.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000405692.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000405692.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000405692.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000426681.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000426681.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000482462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000482462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000482462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000482462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | ENST00000482462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | NM_002607.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | NM_002607.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | NM_002607.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | NM_002607.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | NM_002607.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | NM_002607.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | NM_002607.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | NM_002607.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | NM_002607.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | NM_002607.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | NM_002607.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | NM_002607.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | NM_033023.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | NM_033023.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | NM_033023.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | NM_033023.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | NM_033023.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | NM_033023.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | NM_033023.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | NM_033023.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | NM_033023.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | NM_033023.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009977.hg.1 | NM_033023.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07016420.hg.1 | ENST00000354513.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07016420.hg.1 | ENST00000402802.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07016420.hg.1 | NM_002607.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07016420.hg.1 | NM_033023.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07016420.hg.1 | TC07001073.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0700170965.hg.1 | ENST00000354513.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700170965.hg.1 | ENST00000402802.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700170965.hg.1 | NM_002607.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700170965.hg.1 | NM_033023.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700170965.hg.1 | ENST00000354513.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700170965.hg.1 | ENST00000402802.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700170965.hg.1 | NM_002607.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700170965.hg.1 | NM_033023.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700170965.hg.1 | TC0700009977.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000354513.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000354513.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000354513.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000354513.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000354513.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000354513.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000354513.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000354513.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000354513.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000354513.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000400761.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000400761.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000400761.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000400761.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000400761.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000400761.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000402802.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000402802.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000402802.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000402802.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000402802.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000402802.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000402802.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000402802.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000402802.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000402802.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000405692.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000405692.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000405692.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000405692.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000405692.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000405692.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000426681.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000426681.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000482462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000482462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000482462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000482462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | ENST00000482462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | NM_002607.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | NM_002607.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | NM_002607.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | NM_002607.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | NM_002607.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | NM_002607.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | NM_002607.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | NM_002607.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | NM_002607.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | NM_002607.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | NM_002607.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | NM_002607.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | NM_033023.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | NM_033023.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | NM_033023.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | NM_033023.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | NM_033023.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | NM_033023.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | NM_033023.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | NM_033023.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | NM_033023.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | NM_033023.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009977.hg.1 | NM_033023.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700170964.hg.1 | ENST00000402802.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700170964.hg.1 | NM_002607.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700170964.hg.1 | NM_033023.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700170964.hg.1 | ENST00000402802.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700170964.hg.1 | NM_002607.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700170964.hg.1 | NM_033023.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700170964.hg.1 | TC0700009977.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700170966.hg.1 | ENST00000354513.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700170966.hg.1 | ENST00000400761.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700170966.hg.1 | ENST00000402802.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700170966.hg.1 | NM_002607.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700170966.hg.1 | NM_033023.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700170966.hg.1 | ENST00000354513.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700170966.hg.1 | ENST00000400761.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700170966.hg.1 | ENST00000402802.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700170966.hg.1 | NM_002607.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700170966.hg.1 | NM_033023.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700170966.hg.1 | TC0700009977.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 194326 | - | - |

PDGFA
Genomic Map