Assay Details
| Gene Symbol: LAMA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005560.4 | NP_005551.3 | 39 - 40 | 5281 | 57 | |
| XM_006723796.3 | XP_006723859.1 | 39 - 40 | 5284 | 57 | ||
| XM_006723798.3 | XP_006723861.1 | 39 - 40 | 5287 | 57 | ||
| XM_011528818.2 | XP_011527120.1 | 38 - 39 | 5146 | 57 | ||
| XM_011528819.2 | XP_011527121.1 | 37 - 38 | 5011 | 57 | ||
| GenBank mRNA | AB067494.1 | - | 36 - 37 | 4625 | 57 | |
| AF443072.1 | - | 39 - 40 | 5281 | 57 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20001008.hg.1
Set Membership: |
|

LAMA5
Genomic Map