Assay Details
| Gene Symbol: LAMA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005560.4 | NP_005551.3 | 75 - 76 | 10511 | 100 | |
| XM_006723796.3 | XP_006723859.1 | 75 - 76 | 10529 | 100 | ||
| XM_011528818.2 | XP_011527120.1 | 74 - 75 | 10391 | 100 | ||
| XM_011528819.2 | XP_011527121.1 | 73 - 74 | 10256 | 100 | ||
| GenBank mRNA | AB011105.1 | - | 30 - 31 | 4296 | 100 | |
| AF443072.1 | - | 75 - 76 | 10511 | 100 | ||
| AK074307.1 | - | 13 - 14 | 1826 | 100 | ||
| Z95636.1 | - | 15 - 16 | 2218 | 100 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20001008.hg.1
Set Membership: |
|

LAMA5
Genomic Map