Assay Details
| Gene Symbol: DNAJC13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015268.3 | NP_056083.3 | 39 - 40 | 4605 | 72 | |
| XM_005247245.4 | XP_005247302.1 | 40 - 41 | 4802 | 72 | ||
| XM_017006036.1 | XP_016861525.1 | 26 - 27 | 3062 | 72 | ||
| GenBank mRNA | AB014578.2 | - | 39 - 40 | 4654 | 72 | |
| AK023168.1 | - | 8 - 9 | 1033 | 72 | ||
| AK125330.1 | - | 16 - 17 | 1989 | 72 | ||
| AK127112.1 | - | 23 - 24 | 2693 | 72 | ||
| AY779857.1 | - | 39 - 40 | 4605 | 72 | ||
| BC151246.1 | - | 40 - 41 | 4675 | 9007477 | 72 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000715.hg.1
Set Membership: |
|

DNAJC13
Genomic Map