| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03029204.hg.1 |
ENST00000314261.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03029204.hg.1 |
ENST00000348610.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03029204.hg.1 |
ENST00000358203.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03029204.hg.1 |
ENST00000394085.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03029204.hg.1 |
ENST00000394091.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03029204.hg.1 |
ENST00000394094.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03029204.hg.1 |
ENST00000394095.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03029204.hg.1 |
NM_001077203.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03029204.hg.1 |
NM_020654.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03029204.hg.1 |
TC03001614.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0300188157.hg.1 |
ENST00000314261.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300188157.hg.1 |
ENST00000348610.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300188157.hg.1 |
ENST00000394085.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300188157.hg.1 |
ENST00000394091.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300188157.hg.1 |
ENST00000394094.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300188157.hg.1 |
ENST00000394095.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300188157.hg.1 |
NM_001077203.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300188157.hg.1 |
NM_001282801.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300188157.hg.1 |
NM_001282802.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300188157.hg.1 |
NM_001282803.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300188157.hg.1 |
NM_020654.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300188157.hg.1 |
ENST00000314261.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300188157.hg.1 |
ENST00000348610.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300188157.hg.1 |
ENST00000394085.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300188157.hg.1 |
ENST00000394091.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300188157.hg.1 |
ENST00000394094.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300188157.hg.1 |
ENST00000394095.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300188157.hg.1 |
NM_001077203.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300188157.hg.1 |
NM_001282801.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300188157.hg.1 |
NM_001282802.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300188157.hg.1 |
NM_001282803.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300188157.hg.1 |
NM_020654.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300188157.hg.1 |
TC0300011853.hg.1 |
Clariom D TC |