Assay Details
| Gene Symbol: TRIP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321851.1 | NP_001308780.1 | 5 - 6 | 1033 | 142 | |
| NM_004239.4 | NP_004230.2 | 5 - 6 | 1036 | 142 | ||
| XM_017021788.1 | XP_016877277.1 | 5 - 6 | 1036 | 142 | ||
| GenBank mRNA | AF007217.1 | - | 5 - 6 | 1013 | 142 | |
| AK307489.1 | - | 5 - 6 | 1036 | 142 | ||
| BC146845.1 | - | 5 - 6 | 918 | 9056773 | 142 | |
| Y12490.1 | - | 5 - 6 | 1018 | 142 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14001444.hg
Additional Information:
This assay is in the 5'UTR region of the XM_017021788.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14016163.hg.1 | ENST00000267622.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14016163.hg.1 | ENST00000542257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14016163.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14016163.hg.1 | TC14001444.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000555105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000555105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000555105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000555105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000555105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000555516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000555516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000555516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000555516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000555516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000557017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000557017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000557017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000557017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000557017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000557017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | ENST00000557017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400183156.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183156.hg.1 | ENST00000555516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183156.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400183156.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183156.hg.1 | ENST00000555516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183156.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400183156.hg.1 | TC1400010778.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000554357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000555105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000555105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000555105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000555105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000555105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000555516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000555516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000555516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000555516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000555516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000557017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000557017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000557017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000557017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000557017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000557017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | ENST00000557017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010778.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400183155.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183155.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400183155.hg.1 | ENST00000267622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183155.hg.1 | NM_004239.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400183155.hg.1 | TC1400010778.hg.1 | Clariom D TC |

TRIP11
Genomic Map