Assay Details
| Gene Symbol: MIPEP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005932.3 | NP_005923.2 | 9 - 10 | 1178 | 155 | |
| XM_011535097.2 | XP_011533399.1 | 9 - 10 | 941 | 155 | ||
| XM_011535098.2 | XP_011533400.1 | 9 - 10 | 1178 | 155 | ||
| GenBank mRNA | AK225290.1 | - | 9 - 10 | 1128 | 155 | |
| AK291923.1 | - | 9 - 10 | 1054 | 155 | ||
| BC009934.2 | - | 9 - 10 | 1150 | 4053687 | 155 | |
| U80034.1 | - | 9 - 10 | 155 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000484.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR13005467.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005467.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005467.hg.1 | TC13000484.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005466.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005466.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005466.hg.1 | TC13000484.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000433710.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000433710.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000433710.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000433710.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000433710.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000464194.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000464194.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000464194.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000464194.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000469167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000469167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000469167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000469167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000469167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000494139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000494139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000494139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000494139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000494139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | ENST00000494139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005468.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005468.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005468.hg.1 | TC13000484.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012292.hg.1 | ENST00000413533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012292.hg.1 | ENST00000413533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012292.hg.1 | ENST00000413533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012292.hg.1 | ENST00000413533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000433710.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000433710.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000433710.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000433710.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000433710.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000464194.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000464194.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000464194.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000464194.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000469167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000469167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000469167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000469167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000469167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000494139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000494139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000494139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000494139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000494139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | ENST00000494139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008338.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300156289.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300156289.hg.1 | ENST00000494139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300156289.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300156289.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300156289.hg.1 | ENST00000494139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300156289.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300156289.hg.1 | TC1300008338.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300156290.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300156290.hg.1 | ENST00000494139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300156290.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300156290.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300156290.hg.1 | ENST00000494139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300156290.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300156290.hg.1 | TC1300008338.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300156291.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300156291.hg.1 | ENST00000494139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300156291.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300156291.hg.1 | ENST00000382172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300156291.hg.1 | ENST00000494139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300156291.hg.1 | NM_005932.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300156291.hg.1 | TC1300008338.hg.1 | Clariom D TC |

MIPEP
Genomic Map