Assay Details
| Gene Symbol: PPP1R2P9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002191.2 | 1 | 673 | 145 | ||
| GenBank mRNA | AB044137.1 | - | 1 | 673 | 145 | |
| BC056673.1 | - | 1 | 603 | 145 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | calcium/calmodulin dependent serine protein kinase | CASK |
CAGH39, CAMGUK, CMG, FGS4, LIN2, MICPCH, MRXSNA, TNRC8 |
-854.3kb | Sense | Chr. X - 41514934 - 41923525 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101927501 | LOC101927501 | 398.8kb | Anti-sense | Chr. X - 43176994 - 43226598 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17337199 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32428847 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32428867 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32428878 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11723046 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32428864 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11723028 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11723001 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11723050 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32428877 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32428897 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32428853 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32428869 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11723021 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32428894 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32428879 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11723006 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32428860 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11723057 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32428840 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32428891 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11723015 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11723045 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11723054 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32428880 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11723042 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32428896 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11723058 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32428875 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17337197 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32428898 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11723022 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32428846 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32428889 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32428854 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11723029 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11723032 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32428863 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32428858 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32428844 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11723044 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32428899 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32428868 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32428873 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11723048 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11723041 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32428839 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32428852 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11723023 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32428862 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11723043 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P34007 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P34007 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 6074 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P122947 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P122947 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 44710 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P34007 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P34007 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11723041 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32428879 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32428875 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11723047 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32428880 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11723045 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32428878 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11723028 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11723029 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32428873 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11723046 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32428891 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11723048 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32428890 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11723044 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32428889 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11723050 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11723032 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11723043 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32428869 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11723053 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32428880 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11723047 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32428889 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32428875 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11723050 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11723044 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32428869 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11723046 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11723053 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32428873 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11723048 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32428879 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11723029 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11723041 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32428890 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11723045 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11723043 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11723028 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32428878 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11723032 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32428891 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P34007 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P122947 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P122947 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P122947 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P34007 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 41128 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 96201 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11723000 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11723060 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11723013 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11722999 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11723008 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11723009 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11723017 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11723048 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11723055 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11723024 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11723043 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11723010 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11723019 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11723047 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11723050 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11723039 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11723001 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11723004 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11723058 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11723021 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17337195 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11723031 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11723034 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11723025 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11723046 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11723006 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11723037 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11723040 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11723007 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11723044 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17337199 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11723015 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11723028 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NR_002191_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8172199 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8172198 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8172200 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8172201 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8172197 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 207377_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11745031_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11745031_at | - | - |
| Affymetrix Human Clariom D Assay | PSR0X00164756.hg.1 | NR_002191.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00164756.hg.1 | NR_002191.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00164756.hg.1 | TC0X00009475.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 207377_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 207377_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NR_002191_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw543087 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207377_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207377_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207377_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207377_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207377_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207377_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207377_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207377_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207377_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207377_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207377_at11 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A039775 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A039774 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A039774 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A039775 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 190425 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_2204142 | AB044137.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2204142 | BC056673.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2204142 | NR_002191.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1766057 | - | - |
| HumanHT-12 V4.0 expression beadchip | ILMN_2204142 | AB044137.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2204142 | BC056673.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2204142 | NR_002191.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2204142 | AB044137.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2204142 | BC056673.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2204142 | NR_002191.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2204142 | AB044137.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2204142 | BC056673.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2204142 | NR_002191.2 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1766057 | - | - |
| Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_5701 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_5701 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2204142 | AB044137.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2204142 | BC056673.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2204142 | NR_002191.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2204142 | AB044137.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2204142 | BC056673.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2204142 | NR_002191.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2204142 | AB044137.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2204142 | BC056673.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2204142 | NR_002191.2 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1766057 | - | - |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2204142 | AB044137.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2204142 | BC056673.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2204142 | NR_002191.2 | human_transcriptome |

PPP1R2P9
Genomic Map