Assay Details
| Gene Symbol: ISG20L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005245516.4 | XP_005245573.1 | 1 - 2 | 53 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005245516.4 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000469074.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000469074.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000469074.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000469074.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000470713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000470713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000470713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000470713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000470713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000472824.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000472824.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000472824.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000472824.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000472824.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000496538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000496538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000496538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000496538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000496538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000469074.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000469074.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000469074.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000469074.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000470713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000470713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000470713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000470713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000470713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000472824.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000472824.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000472824.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000472824.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000472824.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000496538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000496538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000496538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000496538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000496538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | NM_001303095.1 | Refseq |

ISG20L2
Genomic Map