Assay Details
| Gene Symbol: ISG20L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001303095.1 | NP_001290024.1 | 3 - 4 | 1150 | 74 | |
| XM_005245516.4 | XP_005245573.1 | 3 - 4 | 1118 | 74 | ||
| XM_005245518.4 | XP_005245575.1 | 2 - 3 | 665 | 74 | ||
| XM_006711560.3 | XP_006711623.1 | 3 - 4 | 1288 | 74 | ||
| XM_006711561.3 | XP_006711624.1 | 3 - 4 | 1177 | 74 | ||
| GenBank mRNA | AK021901.1 | - | 2 - 3 | 330 | 74 | |
| AK022733.1 | - | 2 - 3 | 1208 | 74 | ||
| AK095697.1 | - | 2 - 3 | 1730 | 74 | ||
| AL096734.1 | - | 1 - 2 | 74 | |||
| AY391837.1 | - | 2 - 3 | 1136 | 74 | ||
| BC000575.2 | - | 3 - 4 | 1130 | 3162193 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000469074.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000469074.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000469074.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000469074.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000470713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000470713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000470713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000470713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000470713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000472824.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000472824.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000472824.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000472824.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000472824.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000496538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000496538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000496538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000496538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | ENST00000496538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221548.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221548.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221548.hg.1 | ENST00000469074.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221548.hg.1 | ENST00000470713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221548.hg.1 | ENST00000496538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221548.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221548.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221548.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221548.hg.1 | ENST00000469074.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221548.hg.1 | ENST00000470713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221548.hg.1 | ENST00000496538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221548.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221548.hg.1 | TC0100016020.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100221553.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221553.hg.1 | ENST00000470713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221553.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221553.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221553.hg.1 | ENST00000470713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221553.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221553.hg.1 | TC0100016020.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100221547.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221547.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221547.hg.1 | ENST00000470713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221547.hg.1 | ENST00000496538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221547.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221547.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221547.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221547.hg.1 | ENST00000470713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221547.hg.1 | ENST00000496538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221547.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221547.hg.1 | TC0100016020.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100221541.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221541.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221541.hg.1 | ENST00000472824.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221541.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221541.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221541.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221541.hg.1 | ENST00000472824.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221541.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221541.hg.1 | TC0100016020.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100221542.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221542.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221542.hg.1 | ENST00000472824.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221542.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221542.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221542.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221542.hg.1 | ENST00000472824.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221542.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221542.hg.1 | TC0100016020.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100221544.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221544.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221544.hg.1 | ENST00000472824.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221544.hg.1 | ENST00000496538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221544.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221544.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221544.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221544.hg.1 | ENST00000472824.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221544.hg.1 | ENST00000496538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221544.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221544.hg.1 | TC0100016020.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100221546.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221546.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221546.hg.1 | ENST00000496538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221546.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221546.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221546.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221546.hg.1 | ENST00000496538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221546.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221546.hg.1 | TC0100016020.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100221549.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221549.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221549.hg.1 | ENST00000469074.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221549.hg.1 | ENST00000470713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221549.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221549.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221549.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221549.hg.1 | ENST00000469074.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221549.hg.1 | ENST00000470713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221549.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221549.hg.1 | TC0100016020.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000469074.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000469074.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000469074.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000469074.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000470713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000470713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000470713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000470713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000470713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000472824.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000472824.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000472824.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000472824.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000472824.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000496538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000496538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000496538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000496538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | ENST00000496538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016020.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221550.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221550.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221550.hg.1 | ENST00000469074.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221550.hg.1 | ENST00000470713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221550.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221550.hg.1 | ENST00000313146.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221550.hg.1 | ENST00000368219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221550.hg.1 | ENST00000469074.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221550.hg.1 | ENST00000470713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221550.hg.1 | NM_001303095.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221550.hg.1 | TC0100016020.hg.1 | Clariom D TC |

ISG20L2
Genomic Map