Assay Details
| Gene Symbol: CDC42SE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038707.1 | NP_001033796.1 | 4 - 5 | 730 | 125 | |
| NM_020239.3 | NP_064624.1 | 3 - 4 | 594 | 125 | ||
| XM_017001847.1 | XP_016857336.1 | 4 - 5 | 674 | 125 | ||
| GenBank mRNA | AF187845.2 | - | 3 - 4 | 519 | 125 | |
| AF286041.1 | - | 2 - 3 | 154 | 125 | ||
| AF286592.1 | - | 2 - 3 | 1071 | 125 | ||
| BC012796.2 | - | 2 - 3 | 338 | 3924598 | 125 | |
| BC041604.1 | - | 3 - 4 | 556 | 5473010 | 125 | |
| BX339337.2 | - | 4 - 5 | 642 | 124 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003218.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01049453.hg.1 | ENST00000357235.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01049453.hg.1 | ENST00000483763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01049453.hg.1 | ENST00000540998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01049453.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01049453.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01049453.hg.1 | TC01003218.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100243097.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100243097.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100243097.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100243097.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100243097.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100243097.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100243097.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100243097.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100243097.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100243097.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100243097.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100243097.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100243097.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100243097.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100243097.hg.1 | TC0100018486.hg.1 | Clariom D TC |

CDC42SE1
Genomic Map