Assay Details
| Gene Symbol: STYK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018423.2 | NP_060893.2 | 2 - 3 | 453 | 73 | |
| XM_011520736.1 | XP_011519038.1 | 3 - 4 | 256 | 73 | ||
| XM_011520737.2 | XP_011519039.1 | 2 - 3 | 293 | 73 | ||
| XM_011520738.2 | XP_011519040.1 | 2 - 3 | 448 | 73 | ||
| XM_017019579.1 | XP_016875068.1 | 2 - 3 | 261 | 73 | ||
| GenBank mRNA | AF251059.1 | - | 2 - 3 | 453 | 73 | |
| AK313906.1 | - | 2 - 3 | 188 | 73 | ||
| AL353940.1 | - | 2 - 3 | 201 | 73 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001226.hg
Additional Information:
This assay is in the 5'UTR region of the AF251059.1,AK313906.1,AL353940.1,NM_018423.2,XM_011520736.1,XM_011520737.2,XM_011520738.2,XM_017019579.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transmembrane receptor protein tyrosine kinase signaling pathway
cell migration
cell differentiation
peptidyl-tyrosine autophosphorylation
regulation of cell proliferation
innate immune response
non-membrane spanning protein tyrosine kinase activity
receptor binding
protein binding
ATP binding
transmembrane receptor protein tyrosine kinase signaling pathway
cell migration
cell differentiation
peptidyl-tyrosine autophosphorylation
regulation of cell proliferation
innate immune response
non-membrane spanning protein tyrosine kinase activity
receptor binding
protein binding
ATP binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12016176.hg.1 | ENST00000075503.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016176.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016176.hg.1 | TC12001226.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000535345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000535345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000535345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000535345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000535345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000535345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000535345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000538867.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000538867.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000538867.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000538867.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000538867.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000538867.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000538867.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000541561.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000541561.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000541561.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000541561.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000541561.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000541561.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000542562.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000542562.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000542562.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000542562.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000542562.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000542562.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000542562.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000542924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | ENST00000542924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016177.hg.1 | ENST00000075503.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016177.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016177.hg.1 | TC12001226.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000535345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000535345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000535345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000535345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000535345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000535345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000535345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000538867.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000538867.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000538867.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000538867.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000538867.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000538867.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000538867.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000541561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000541561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000541561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000541561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000541561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000541561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000542562.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000542562.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000542562.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000542562.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000542562.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000542562.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000542562.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000542924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | ENST00000542924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009912.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200174825.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174825.hg.1 | ENST00000535345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174825.hg.1 | ENST00000538867.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174825.hg.1 | ENST00000541561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174825.hg.1 | ENST00000542562.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174825.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200174825.hg.1 | ENST00000075503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174825.hg.1 | ENST00000535345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174825.hg.1 | ENST00000538867.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174825.hg.1 | ENST00000541561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174825.hg.1 | ENST00000542562.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174825.hg.1 | NM_018423.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200174825.hg.1 | TC1200009912.hg.1 | Clariom D TC |

STYK1
Genomic Map