| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012074.hg.1 |
ENST00000340581.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012074.hg.1 |
ENST00000354791.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012074.hg.1 |
ENST00000356606.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012074.hg.1 |
ENST00000357544.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012074.hg.1 |
ENST00000357991.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012074.hg.1 |
ENST00000360279.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012074.hg.1 |
ENST00000379869.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012074.hg.1 |
ENST00000379873.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012074.hg.1 |
ENST00000379876.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012074.hg.1 |
ENST00000379878.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012074.hg.1 |
ENST00000397917.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012074.hg.1 |
NM_001079858.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012074.hg.1 |
NM_001079859.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012074.hg.1 |
NM_001079860.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012074.hg.1 |
NM_001184833.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012074.hg.1 |
NM_001184834.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012074.hg.1 |
NM_001184835.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012074.hg.1 |
NM_001184836.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012074.hg.1 |
NM_001184837.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012074.hg.1 |
NM_005756.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012074.hg.1 |
TC0X000901.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
ENST00000340581.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
ENST00000354791.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
ENST00000356606.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
ENST00000357544.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
ENST00000357991.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
ENST00000360279.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
ENST00000379869.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
ENST00000379873.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
ENST00000379876.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
ENST00000379878.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
ENST00000479496.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
NM_001079858.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
NM_001079859.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
NM_001079860.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
NM_001184833.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
NM_001184834.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
NM_001184835.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
NM_001184836.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
NM_001184837.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
NM_005756.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
ENST00000340581.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
ENST00000354791.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
ENST00000356606.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
ENST00000379869.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
ENST00000357544.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
ENST00000357991.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
ENST00000360279.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
ENST00000379873.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
ENST00000379876.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
ENST00000379878.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
ENST00000479496.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
NM_001079858.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
NM_001079859.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
NM_001079860.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
NM_001184833.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
NM_001184834.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
NM_001184835.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
NM_001184836.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
NM_001184837.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
NM_005756.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163116.hg.1 |
TC0X00009199.hg.1 |
Clariom D TC |