Assay Details
| Gene Symbol: ADAM22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001324417.1 | NP_001311346.1 | 25 - 27 | 2608 | 137 | |
| NM_001324421.1 | NP_001311350.1 | 25 - 27 | 2611 | 137 | ||
| NM_016351.5 | NP_057435.2 | 25 - 27 | 2611 | 137 | ||
| NM_021721.4 | NP_068367.1 | 25 - 27 | 2611 | 137 | ||
| NM_021722.5 | NP_068368.2 | 25 - 27 | 2611 | 137 | ||
| XM_011516323.1 | XP_011514625.1 | 25 - 27 | 2503 | 137 | ||
| XM_017012329.1 | XP_016867818.1 | 26 - 28 | 2554 | 137 | ||
| XM_017012331.1 | XP_016867820.1 | 25 - 27 | 2584 | 137 | ||
| XM_017012334.1 | XP_016867823.1 | 26 - 28 | 2554 | 137 | ||
| XM_017012335.1 | XP_016867824.1 | 26 - 28 | 2554 | 137 | ||
| XM_017012336.1 | XP_016867825.1 | 25 - 27 | 2503 | 137 | ||
| XM_017012337.1 | XP_016867826.1 | 26 - 28 | 2554 | 137 | ||
| XM_017012339.1 | XP_016867828.1 | 26 - 28 | 2554 | 137 | ||
| XM_017012341.1 | XP_016867830.1 | 24 - 26 | 2231 | 137 | ||
| GenBank mRNA | AF073291.2 | - | 25 - 27 | 2334 | 137 | |
| AF155381.1 | - | 25 - 27 | 2334 | 137 | ||
| AK291010.1 | - | 25 - 27 | 2458 | 137 | ||
| BC125118.1 | - | 25 - 27 | 2365 | 40118904 | 137 | |
More Information
Set Membership: |
|

ADAM22
Genomic Map