Assay Details
| Gene Symbol: SAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027783.2 | 4 - 5 | 505 | 135 | ||
| GenBank mRNA | AL050290.1 | - | 4 - 5 | 466 | 135 | |
| AY841998.1 | - | 4 - 5 | 312 | 135 | ||
| CD638929.1 | - | 4 - 5 | 493 | 135 | ||
More Information
Updated Mapping Information
Set Membership: |
|

SAT1
Genomic Map