Assay Details
| Gene Symbol: LRP2BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018409.3 | NP_060879.2 | 1 - 2 | 917 | 102 | |
| XM_006714260.3 | XP_006714323.1 | 2 - 3 | 1247 | 102 | ||
| XM_017008410.1 | XP_016863899.1 | 2 - 3 | 2130 | 102 | ||
| XM_017008411.1 | XP_016863900.1 | 1 - 2 | 738 | 102 | ||
| GenBank mRNA | AB037746.1 | - | 1 - 2 | 917 | 102 | |
| AK126913.1 | - | 2 - 3 | 1248 | 102 | ||
| BC150161.1 | - | 1 - 2 | 424 | 40126591 | 102 | |
| BC152444.1 | - | 1 - 2 | 917 | 8862572 | 102 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001815.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04024769.hg.1 | ENST00000328559.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024769.hg.1 | ENST00000362004.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024769.hg.1 | ENST00000510776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024769.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024769.hg.1 | TC04001815.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024762.hg.1 | ENST00000328559.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024762.hg.1 | ENST00000362004.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024762.hg.1 | ENST00000505916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024762.hg.1 | ENST00000510776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024762.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024762.hg.1 | TC04001815.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024768.hg.1 | ENST00000328559.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024768.hg.1 | ENST00000362004.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024768.hg.1 | ENST00000510776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024768.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024768.hg.1 | TC04001815.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000511404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000511404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000511404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000511404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | ENST00000511404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013001.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024767.hg.1 | ENST00000328559.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024767.hg.1 | ENST00000362004.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024767.hg.1 | ENST00000510776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024767.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024767.hg.1 | TC04001815.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024766.hg.1 | ENST00000328559.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024766.hg.1 | ENST00000362004.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024766.hg.1 | ENST00000505916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024766.hg.1 | ENST00000510776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024766.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024766.hg.1 | TC04001815.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024770.hg.1 | ENST00000328559.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024770.hg.1 | ENST00000362004.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024770.hg.1 | ENST00000510776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024770.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024770.hg.1 | TC04001815.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0400191514.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191514.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191514.hg.1 | ENST00000511404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191514.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191514.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191514.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191514.hg.1 | ENST00000511404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191514.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191514.hg.1 | TC0400013001.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400191516.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191516.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191516.hg.1 | ENST00000511404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191516.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191516.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191516.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191516.hg.1 | ENST00000511404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191516.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191516.hg.1 | TC0400013001.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400191519.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191519.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191519.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191519.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191519.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191519.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191519.hg.1 | TC0400013001.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400191520.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191520.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191520.hg.1 | ENST00000511404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191520.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191520.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191520.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191520.hg.1 | ENST00000511404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191520.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191520.hg.1 | TC0400013001.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400191521.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191521.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191521.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191521.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191521.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191521.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191521.hg.1 | TC0400013001.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000511404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000511404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000511404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000511404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | ENST00000511404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400013001.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191518.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191518.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191518.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191518.hg.1 | ENST00000328559.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191518.hg.1 | ENST00000510776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191518.hg.1 | NM_018409.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191518.hg.1 | TC0400013001.hg.1 | Clariom D TC |

LRP2BP
Genomic Map