Assay Details
| Gene Symbol: IRAK1BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001010844.3 | NP_001010844.1 | 4 | 2927 | 73 | |
| GenBank mRNA | AK096861.1 | - | 1 | 1599 | 73 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | meiotic double-stranded break formation protein 4 | MEI4 | -971.0kb | Sense | Chr. 6 - 77650274 - 77927045 on NCBI Build GRCh38 | |
| Downstream | pleckstrin homology domain interacting protein | PHIP |
BRWD2, DCAF14, WDR11, ndrp |
33.5kb | Anti-sense | Chr. 6 - 78934419 - 79078294 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

IRAK1BP1
Genomic Map