Assay Details
| Gene Symbol: PPOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011509668.2 | XP_011507970.1 | 1 - 2 | 70 | 110 | |
| XM_011509679.1 | XP_011507981.1 | 1 - 2 | 79 | 110 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011509668.2, XM_011509679.1 transcripts.
Set Membership: |
|

PPOX
Genomic Map