Assay Details
| Gene Symbol: PPM1J | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005167.5 | NP_005158.5 | 2 - 3 | 469 | 79 | |
| GenBank mRNA | AK093270.1 | - | 2 - 3 | 449 | 79 | |
| AL133644.1 | - | 2 - 3 | 1350 | 79 | ||
| BC011803.2 | - | 2 - 3 | 393 | 79 | ||
| BC064628.1 | - | 2 - 3 | 396 | 79 | ||
| BC073828.1 | - | 2 - 3 | 461 | 5244363 | 79 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01006276.hg
Additional Information:
This assay is in the 5'UTR region of the AL133644.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000486709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000486709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000486709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000486709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046620.hg.1 | ENST00000309276.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046620.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046620.hg.1 | TC01006276.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046622.hg.1 | ENST00000309276.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046622.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046622.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046622.hg.1 | TC01006276.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046621.hg.1 | ENST00000309276.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046621.hg.1 | ENST00000359994.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046621.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046621.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046621.hg.1 | TC01006276.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100242620.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242620.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242620.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242620.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100242620.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242620.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242620.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242620.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100242620.hg.1 | TC0100018464.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000486709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000486709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000486709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000486709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100242621.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242621.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242621.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242621.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242621.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100242621.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242621.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242621.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242621.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242621.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100242621.hg.1 | TC0100018464.hg.1 | Clariom D TC |

PPM1J
Genomic Map