Assay Details
| Gene Symbol: PPM1J | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005167.5 | NP_005158.5 | 7 - 8 | 1139 | 69 | |
| GenBank mRNA | AB451327.1 | - | 6 - 7 | 494 | 69 | |
| AB451471.1 | - | 6 - 7 | 494 | 69 | ||
| AK093270.1 | - | 7 - 8 | 1119 | 69 | ||
| AL133644.1 | - | 8 - 9 | 1903 | 69 | ||
| BC011803.2 | - | 7 - 8 | 1063 | 69 | ||
| BC064628.1 | - | 7 - 8 | 1159 | 69 | ||
| BC073828.1 | - | 7 - 8 | 1131 | 5244363 | 69 | |
More Information
Set Membership: |
|

PPM1J
Genomic Map