Assay Details
| Gene Symbol: TAPBPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018009.4 | NP_060479.3 | 2 - 3 | 530 | 88 | |
| XM_005253700.3 | XP_005253757.1 | 2 - 3 | 530 | 88 | ||
| XM_017019545.1 | XP_016875034.1 | 2 - 3 | 678 | 88 | ||
| XM_017019546.1 | XP_016875035.1 | 2 - 3 | 358 | 88 | ||
| GenBank mRNA | AK001005.1 | - | 2 - 3 | 434 | 88 | |
| AK002056.2 | - | 2 - 3 | 418 | 88 | ||
| AK098035.1 | - | 2 - 3 | 617 | 88 | ||
| AK222956.1 | - | 2 - 3 | 418 | 88 | ||
| BC015017.2 | - | 2 - 3 | 457 | 3916213 | 88 | |
| CR997450.1 | - | 2 - 3 | 412 | 88 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000081.hg
Additional Information:
This assay is in the 5'UTR region of the XM_017019545.1,XM_017019546.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12000842.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12000842.hg.1 | ENST00000544021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12000842.hg.1 | ENST00000545700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12000842.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12000842.hg.1 | TC12000081.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000542160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000542160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000542160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000542160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000542160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000542160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000542160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000542160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000542160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000542160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200198874.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200198874.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200198874.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200198874.hg.1 | ENST00000544826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200198874.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200198874.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200198874.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200198874.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200198874.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200198874.hg.1 | ENST00000544826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200198874.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200198874.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200198874.hg.1 | TC1200012584.hg.1 | Clariom D TC |

TAPBPL
Genomic Map