Assay Details
| Gene Symbol: TAPBPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK098035.1 | - | 4 - 5 | 1032 | 57 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000542160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000542160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000542160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000542160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000542160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000544826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000266556.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000542160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000542160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000542160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000542160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000542160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000543567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544021.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000544826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | ENST00000545700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012584.hg.1 | NM_018009.4 | Refseq |

TAPBPL
Genomic Map