Assay Details
| Gene Symbol: PRDM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012406.3 | NP_036538.3 | 4 - 5 | 769 | 68 | |
| XM_005268593.1 | XP_005268650.1 | 4 - 5 | 767 | 68 | ||
| GenBank mRNA | AF144757.2 | - | 3 - 4 | 454 | 68 | |
| AK056378.1 | - | 4 - 5 | 636 | 68 | ||
| AL133083.1 | - | 1 - 2 | 104 | 68 | ||
| BC035581.1 | - | 3 - 4 | 570 | 5587364 | 68 | |
| BC068562.1 | - | 4 - 5 | 702 | 68 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001928.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
signal transduction
cell proliferation
negative regulation of cell growth
cellular response to nutrient
histone H4-R3 methylation
cellular response to growth factor stimulus
DNA binding
death receptor binding
protein binding
methyltransferase activity
zinc ion binding
histone methyltransferase binding
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
signal transduction
cell proliferation
negative regulation of cell growth
cellular response to nutrient
histone H4-R3 methylation
cellular response to growth factor stimulus
DNA binding
death receptor binding
protein binding
methyltransferase activity
zinc ion binding
histone methyltransferase binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12025116.hg.1 | ENST00000228437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025116.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025116.hg.1 | TC12001928.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000547188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000547188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000547188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000547188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000547188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000547268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000547268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000547268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000547268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000547268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000547268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000549416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000549416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000549416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000549416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191597.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191597.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191597.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191597.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191597.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191597.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191597.hg.1 | TC1200011820.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000547188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000547188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000547188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000547188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000547188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000547268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000547268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000547268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000547268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000547268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000547268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000549416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000549416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000549416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000549416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |

PRDM4
Genomic Map