| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11020959.hg.1 |
ENST00000278400.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11020959.hg.1 |
ENST00000344743.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11020959.hg.1 |
ENST00000529732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11020959.hg.1 |
NM_005838.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11020959.hg.1 |
NM_201648.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11020959.hg.1 |
TC11001813.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000278400.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000278400.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000278400.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000278400.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000278400.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000278400.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000278400.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000278400.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000278400.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000344743.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000344743.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000344743.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000344743.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000344743.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000344743.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000344743.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000344743.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000344743.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000344743.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000344743.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000529732.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000529732.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000529732.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000529732.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000529732.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000529732.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000529732.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000529732.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000529732.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000531084.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000531084.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000531084.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000586098.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000586098.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000586098.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000611865.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000611865.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000611865.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000611865.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000611865.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000611865.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000611865.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000611865.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000611865.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
ENST00000611865.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
NM_005838.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
NM_005838.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
NM_005838.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
NM_005838.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
NM_005838.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
NM_005838.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
NM_005838.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
NM_005838.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
NM_005838.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
NM_201648.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
NM_201648.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
NM_201648.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
NM_201648.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
NM_201648.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
NM_201648.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
NM_201648.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
NM_201648.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
NM_201648.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
NM_201648.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
NM_201648.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010948.hg.1 |
NM_201648.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100182646.hg.1 |
ENST00000278400.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100182646.hg.1 |
ENST00000344743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100182646.hg.1 |
ENST00000529732.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100182646.hg.1 |
ENST00000611865.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100182646.hg.1 |
NM_005838.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100182646.hg.1 |
NM_201648.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100182646.hg.1 |
ENST00000278400.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100182646.hg.1 |
ENST00000344743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100182646.hg.1 |
ENST00000529732.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100182646.hg.1 |
ENST00000611865.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100182646.hg.1 |
NM_005838.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100182646.hg.1 |
NM_201648.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100182646.hg.1 |
TC1100010948.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100182645.hg.1 |
ENST00000278400.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100182645.hg.1 |
ENST00000344743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100182645.hg.1 |
ENST00000529732.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100182645.hg.1 |
ENST00000586098.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100182645.hg.1 |
ENST00000611865.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100182645.hg.1 |
NM_005838.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100182645.hg.1 |
NM_201648.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100182645.hg.1 |
ENST00000278400.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100182645.hg.1 |
ENST00000344743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100182645.hg.1 |
ENST00000529732.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100182645.hg.1 |
ENST00000586098.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100182645.hg.1 |
ENST00000611865.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100182645.hg.1 |
NM_005838.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100182645.hg.1 |
NM_201648.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100182645.hg.1 |
TC1100010948.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000278400.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000278400.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000278400.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000278400.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000278400.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000278400.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000278400.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000278400.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000278400.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000344743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000344743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000344743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000344743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000344743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000344743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000344743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000344743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000344743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000344743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000344743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000529732.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000529732.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000529732.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000529732.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000529732.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000529732.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000529732.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000529732.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000529732.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000531084.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000531084.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000531084.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000586098.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000586098.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000586098.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000611865.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000611865.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000611865.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000611865.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000611865.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000611865.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000611865.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000611865.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000611865.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
ENST00000611865.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
NM_005838.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
NM_005838.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
NM_005838.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
NM_005838.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
NM_005838.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
NM_005838.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
NM_005838.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
NM_005838.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
NM_005838.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
NM_201648.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
NM_201648.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
NM_201648.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
NM_201648.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
NM_201648.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
NM_201648.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
NM_201648.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
NM_201648.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
NM_201648.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
NM_201648.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
NM_201648.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010948.hg.1 |
NM_201648.2 |
Refseq |