Assay Details
| Gene Symbol: PRPF19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014502.4 | NP_055317.1 | 8 - 9 | 845 | 78 | |
| GenBank mRNA | AJ131186.1 | - | 8 - 9 | 845 | 78 | |
| BC008719.2 | - | 8 - 9 | 840 | 2961655 | 78 | |
| BC018665.2 | - | 8 - 9 | 840 | 2961655 | 78 | |
| BC018698.2 | - | 8 - 9 | 840 | 2961655 | 78 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001844.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
spliceosomal tri-snRNP complex assembly
spliceosomal complex assembly
mRNA splicing, via spliceosome
inner cell mass cell proliferation
transcription-coupled nucleotide-excision repair
double-strand break repair via nonhomologous end joining
lipid biosynthetic process
proteasomal protein catabolic process
cellular protein localization
negative regulation of neuron differentiation
positive regulation of mRNA splicing, via spliceosome
positive regulation of astrocyte differentiation
protein K63-linked ubiquitination
signal transduction involved in DNA damage checkpoint
protein binding
ligase activity
ubiquitin-ubiquitin ligase activity
identical protein binding
ubiquitin protein ligase activity
protein polyubiquitination
spliceosomal tri-snRNP complex assembly
spliceosomal complex assembly
mRNA splicing, via spliceosome
inner cell mass cell proliferation
transcription-coupled nucleotide-excision repair
double-strand break repair via nonhomologous end joining
lipid biosynthetic process
proteasomal protein catabolic process
cellular protein localization
negative regulation of neuron differentiation
positive regulation of mRNA splicing, via spliceosome
positive regulation of astrocyte differentiation
protein K63-linked ubiquitination
signal transduction involved in DNA damage checkpoint
protein binding
ligase activity
ubiquitin-ubiquitin ligase activity
identical protein binding
ubiquitin protein ligase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000535326.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000535326.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000535326.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000535326.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000535326.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000535326.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000535326.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000535326.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000539180.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000539180.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000539180.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000539960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000539960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000539960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000539960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000539960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000539960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000539960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000539960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000540473.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000540473.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000540473.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000541371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000541371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000541371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000541371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000541371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000541371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000541371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000541371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000541371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000541371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000541371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000541371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000546152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000546152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000546152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000546152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000546152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000546152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000546152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000546152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | ENST00000546152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11021194.hg.1 | ENST00000227524.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11021194.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11021194.hg.1 | TC11001844.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000535326.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000535326.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000535326.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000535326.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000535326.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000535326.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000535326.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000535326.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000539180.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000539180.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000539180.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000539960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000539960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000539960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000539960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000539960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000539960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000539960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000539960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000540473.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000540473.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000540473.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000541371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000541371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000541371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000541371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000541371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000541371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000541371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000541371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000541371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000541371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000541371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000541371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000546152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000546152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000546152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000546152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000546152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000546152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000546152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000546152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | ENST00000546152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011010.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100183070.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100183070.hg.1 | ENST00000541371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100183070.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100183070.hg.1 | ENST00000227524.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100183070.hg.1 | ENST00000541371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100183070.hg.1 | NM_014502.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100183070.hg.1 | TC1100011010.hg.1 | Clariom D TC |

PRPF19
Genomic Map