Assay Details
| Gene Symbol: MYRF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127392.2 | NP_001120864.1 | 16 - 17 | 2356 | 112 | |
| NM_013279.3 | NP_037411.1 | 16 - 17 | 2256 | 112 | ||
| XM_005274222.1 | XP_005274279.1 | 16 - 17 | 2344 | 112 | ||
| XM_005274223.1 | XP_005274280.1 | 16 - 17 | 2344 | 112 | ||
| XM_005274224.1 | XP_005274281.1 | 16 - 17 | 2344 | 112 | ||
| XM_005274225.1 | XP_005274282.1 | 16 - 17 | 2344 | 112 | ||
| XM_005274226.1 | XP_005274283.1 | 16 - 17 | 2344 | 112 | ||
| XM_005274227.1 | XP_005274284.1 | 16 - 17 | 2344 | 112 | ||
| XM_005274228.1 | XP_005274285.1 | 16 - 17 | 2191 | 112 | ||
| XM_011545234.2 | XP_011543536.1 | 16 - 17 | 2230 | 112 | ||
| GenBank mRNA | AB023171.2 | - | 16 - 17 | 2344 | 112 | |
| AF086762.1 | - | 16 - 17 | 2256 | 112 | ||
| AK295013.1 | - | 4 - 5 | 585 | 112 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000539.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
proteolysis
oligodendrocyte development
central nervous system myelination
positive regulation of myelination
central nervous system myelin maintenance
positive regulation of transcription, DNA-templated
oligodendrocyte differentiation
DNA binding
transcription factor activity, sequence-specific DNA binding
peptidase activity
transcription, DNA-templated
proteolysis
oligodendrocyte development
central nervous system myelination
positive regulation of myelination
central nervous system myelin maintenance
positive regulation of transcription, DNA-templated
oligodendrocyte differentiation
DNA binding
transcription factor activity, sequence-specific DNA binding
peptidase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000536352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000536352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000536352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000536352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000536352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000536352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000536352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000536352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000539361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000539361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000539361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000539361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000539361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000539361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000539361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000539361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000546247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000546247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000546247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000546247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000546247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |

MYRF
Genomic Map