Assay Details
| Gene Symbol: MYRF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127392.2 | NP_001120864.1 | 22 - 23 | 3125 | 119 | |
| XM_005274222.1 | XP_005274279.1 | 22 - 23 | 3116 | 119 | ||
| XM_005274225.1 | XP_005274282.1 | 21 - 22 | 3035 | 119 | ||
| XM_005274226.1 | XP_005274283.1 | 21 - 22 | 3032 | 119 | ||
| XM_005274228.1 | XP_005274285.1 | 22 - 23 | 2963 | 119 | ||
| XM_011545234.2 | XP_011543536.1 | 22 - 23 | 3002 | 119 | ||
| GenBank mRNA | AB023171.2 | - | 22 - 23 | 3113 | 119 | |
More Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
proteolysis
oligodendrocyte development
central nervous system myelination
positive regulation of myelination
central nervous system myelin maintenance
positive regulation of transcription, DNA-templated
oligodendrocyte differentiation
DNA binding
transcription factor activity, sequence-specific DNA binding
peptidase activity
transcription, DNA-templated
proteolysis
oligodendrocyte development
central nervous system myelination
positive regulation of myelination
central nervous system myelin maintenance
positive regulation of transcription, DNA-templated
oligodendrocyte differentiation
DNA binding
transcription factor activity, sequence-specific DNA binding
peptidase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000536352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000536352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000536352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000536352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000536352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000536352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000536352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000536352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000539361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000539361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000539361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000539361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000539361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000539361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000539361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000539361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000546247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000546247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000546247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000546247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000546247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |

MYRF
Genomic Map