| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000265460.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000265460.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000265460.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000265460.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000265460.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000265460.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000265460.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000265460.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000265460.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000265460.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000265460.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000265460.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000265460.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000265460.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000265460.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000265460.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000265460.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000265460.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000265460.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000265460.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000265460.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000265460.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000265460.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000265460.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000265460.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000265460.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000265460.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000265460.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000265460.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000278836.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000278836.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000278836.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000278836.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000278836.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000278836.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000278836.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000278836.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000278836.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000278836.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000278836.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000278836.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000278836.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000278836.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000278836.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000278836.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000278836.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000278836.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000278836.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000278836.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000278836.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000278836.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000278836.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000278836.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000278836.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000278836.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000278836.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000278836.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000278836.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000278836.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000278836.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000389602.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000389602.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000389602.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000389602.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000389602.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000389602.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000389602.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000389602.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000389602.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000389602.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000389602.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000389602.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000389602.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000389602.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000389602.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000389602.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000389602.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000389602.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000536352.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000536352.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000536352.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000536352.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000536352.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000536352.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000536352.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000536352.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000537318.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000537318.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000537318.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000537318.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000537318.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000537318.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000537318.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000537318.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000537318.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000537318.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000537766.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000537766.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000537766.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000537766.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000537766.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000537766.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000539361.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000539361.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000539361.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000539361.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000539361.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000539361.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000539361.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000539361.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000546247.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000546247.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000546247.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000546247.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
ENST00000546247.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_001127392.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_001127392.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_001127392.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_001127392.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_001127392.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_001127392.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_001127392.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_001127392.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_001127392.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_001127392.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_001127392.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_001127392.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_001127392.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_001127392.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_001127392.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_001127392.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_001127392.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_001127392.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_001127392.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_001127392.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_001127392.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_001127392.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_001127392.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_001127392.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_001127392.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_001127392.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_001127392.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_001127392.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_001127392.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_001127392.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_001127392.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_013279.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_013279.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_013279.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_013279.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_013279.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_013279.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_013279.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_013279.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_013279.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_013279.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_013279.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_013279.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_013279.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_013279.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_013279.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_013279.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_013279.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_013279.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_013279.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_013279.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_013279.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_013279.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_013279.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_013279.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_013279.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_013279.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_013279.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_013279.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007819.hg.1 |
NM_013279.3 |
Refseq |